Skip to Content
Merck
All Photos(1)

Key Documents

P1240000

Phenylephrine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Phenylephrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenylephrine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N(C[C@H](O)c1cc(ccc1)O)C

InChI

1S/C9H13NO2/c1-10-6-9(12)7-3-2-4-8(11)5-7/h2-5,9-12H,6H2,1H3/t9-/m0/s1

InChI key

SONNWYBIRXJNDC-VIFPVBQESA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenylephrine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uwe Kirchhefer et al.
The Journal of biological chemistry, 289(1), 163-176 (2013-11-15)
Protein phosphatase 2A (PP2A) is a family of multifunctional serine/threonine phosphatases consisting of a catalytic C, a structural A, and a regulatory B subunit. The substrate and therefore the functional specificity of PP2A are determined by the assembly of the
Ashraf S Habib
Anesthesia and analgesia, 114(2), 377-390 (2011-11-23)
Phenylephrine is effective for the management of spinal anesthesia-induced hypotension in parturients undergoing cesarean delivery under spinal anesthesia. While ephedrine was previously considered the vasopressor of choice in obstetric patients, phenylephrine is increasingly being used. This is largely due to
Hartley C Atkinson et al.
European journal of clinical pharmacology, 71(2), 151-158 (2014-12-06)
Over-the-counter combinations containing acetaminophen and phenylephrine for treatment of the common cold and influenza are widespread, but there are few data about pharmacokinetics of these two drugs used in combination. We aimed to investigate pharmacokinetic interactions between acetaminophen and phenylephrine.
A G Tahiliani et al.
General pharmacology, 13(5), 369-374 (1982-01-01)
To summarize, a large number of studies indicate that the effects of phenylephrine on cardiac contractility are mediated through alpha-adrenoceptors, although not exclusively. There are an equally large number of reports indicating some beta-adrenoceptor stimulant effects of phenylephrine. Of the
Qu Nie et al.
Life sciences, 232, 116662-116662 (2019-07-20)
Vascular endothelial cells act as a selective barrier between circulating blood and vessel wall and play an important role in the occurrence and development of cardiovascular diseases. Astragaloside IV (As-IV) has a protective effect on vascular endothelial cells, but its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service