Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(
R
)-(−)-Phenylephrine hydrochloride
Synonym(s):
(
R
)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (
R
)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride
Linear Formula:
HOC
6
H
4
CH(CH
2
NHCH
3
)OH·HCl
CAS Number:
61-76-7
Molecular Weight:
203.67
EC Number:
200-517-3
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Research Area
Quality Segment
Package Size
Special Grade
analytical standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P6126
(
R
)-(−)-Phenylephrine hydrochloride,
≥99% (titration), powder, α₁-adrenoceptor agonist
Expand
View Pricing
P8155
(
R
)-(−)-Phenylephrine hydrochloride,
analytical standard
Expand
View Pricing