P2307
Polyvinylpyrrolidone
suitable for plant cell culture, average mol wt 10,000
Synonym(s):
PVP, Polyvidone, Povidone
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
Recommended Products
form
powder
Quality Level
mol wt
average mol wt 10,000
technique(s)
cell culture | plant: suitable
viscosity number
12-18(lit.)
SMILES string
C=CN1CCCC1=O
InChI
1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2
InChI key
WHNWPMSKXPGLAX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Design and development of a soluble PDA-Emodin-PVP-MN patch and its anti-obesity effect in rats.: This study presents the development of a microneedle patch containing Polyvinylpyrrolidone (PVP), which enhances the delivery of anti-obesity medication, showcasing PVP′s utility in pharmaceutical formulations to improve drug solubility and efficacy (Wang H et al., 2024).
- Polyvinylpyrrolidone Assisted One-Pot Synthesis of Size-Tunable Cocktail Nanodrug for Multifunctional Combat of Cancer.: This research demonstrates the role of PVP, as a stabilizer and regulator in the synthesis of a nanodrug cocktail, indicating its critical function in nanomedicine for targeting and treating cancer effectively, thereby optimizing therapeutic outcomes (Wang C et al., 2024).
- Biochemical parameters, oxidative stress biomarkers, and anatomopathological changes in Wistar rats treated with 3′-demethoxy-6-O-demethylisoguaiacin and norisoguaiacin.: The study utilizes PVP in biomedical applications, particularly in enhancing the solubility and bioavailability of specific bioactive compounds for therapeutic purposes (Flores Jimenez NG et al., 2024).
- A water-soluble preparation for intravenous administration of isorhamnetin and its pharmacokinetics in rats.: Highlighting PVP′s importance in creating water-soluble drug formulations that facilitate the intravenous delivery of hydrophobic substances, this study explores its application in improving drug delivery systems (Rassu G et al., 2024).
- Activation of the PPAR gamma/NF-kB pathway by A-MPDA@Fe(3)O(4)@PVP via scavenging reactive oxygen species to alleviate hepatic ischemia-reperfusion injury.: This innovative study employs PVP to stabilize nanoparticle systems that actively participate in reducing oxidative stress, illustrating PVP′s potential in therapeutic interventions (Mo D et al., 2024).
Other Notes
Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Katja A Lüders et al.
Glia, 67(4), 634-649 (2019-01-15)
Proteolipid protein (PLP) is the most abundant integral membrane protein in central nervous system (CNS) myelin. Expression of the Plp-gene in oligodendrocytes is not essential for the biosynthesis of myelin membranes but required to prevent axonal pathology. This raises the
Abeer Maher Fayyad et al.
International journal of molecular sciences, 20(7) (2019-04-04)
Obesity is a major risk for diabetes. Brown adipose tissue (BAT) mediates production of heat while white adipose tissue (WAT) function in the storage of fat. Roles of BAT in the treatment of obesity and related disorders warrants more investigation.
Nicole M Sanders et al.
American journal of physiology. Endocrinology and metabolism, 294(5), E853-E860 (2008-03-13)
Selective serotonin reuptake inhibitors (SSRIs) are widely prescribed for patients with comorbid diabetes and depression. Clinical case studies in diabetic patients, however, suggest that SSRI therapy may exacerbate hypoglycemia. We hypothesized that SSRIs might increase the risk of hypoglycemia by
X C Qin et al.
Journal of photochemistry and photobiology. B, Biology, 120, 156-162 (2013-01-30)
Nanographene oxide (NGO), a new type of nanomaterial for anticancer drugs delivery and near-infrared (NIR)-mediated photothermal ablation of tumors, has been used in the combination of photothermal therapy and chemotherapy. Herein, targeted chemo-photothermal therapy based on polyvinylpyrrolidone (PVP) functionalized NGO
Liyun Ren et al.
Nanoscale, 5(6), 2337-2345 (2013-02-09)
PLLA fibrous tissue scaffolds with controlled fiber nanoscale surface roughness are fabricated with a novel centrifugal jet spinning process. The centrifugal jet spinning technique is a highly efficient synthesis method for micron- to nano-sized fibers with a production rate up
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service