Skip to Content
Merck
All Photos(5)

Documents

PVP40

Sigma-Aldrich

Polyvinylpyrrolidone

average mol wt 40,000

Synonym(s):

PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (oragnic)

Quality Level

form

powder

mol wt

average mol wt 40,000

impurities

<5.0% water (Karl Fischer)

viscosity number

29-32(lit.)

solubility

water: 100 mg/mL, clear, colorless to light yellow

application(s)

agriculture

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyvinylpyrrolidone (PVP) can form complexes with phenolics and alkaloids. The complex formation allows removal of phenolics and alkaloids from plant samples, thereby inhibiting any modification of proteins by them and any hindrance they may cause in spectrophotometric determinations of protein content. PVP might also enhances the stability of enzymes.
Polyvinylpyrrolidone (PVP), a multifunctional synthetic polymer, finds utility in life science and biochemical research. This water-soluble polymer is versatile, serving as a stabilizing agent, emulsifier, suspending agent, solubilizing agent, protective colloid, and complexing agent. Its protective colloid properties are particularly valuable in stabilizing proteins, enzymes, and other biological substances.

Application

Polyvinylpyrrolidone has been used in the purification of glutamine synthetases from plant samples. It has been used to block the PVDF (polyvinylidene difluoride) membrane and to avoid nonspecific binding.

Features and Benefits

  • Highly versatile surfactant for your life science and biochemical research
  • Excellent Solubilizing agent and Complexing agent

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural analysis of N- and O-glycans released from glycoproteins.
Jensen PH et al.
Nature Protocols, 7, 1299-1299 (2012)
Glutamine Synthetases of Higher Plants : Evidence for a Specific Isoform Content Related to Their Possible Physiological Role and Their Compartmentation within the Leaf.
McNally SF et al.
Plant Physiology, 72, 22-22 (1983)
Angiotensin II inhibits insulin signaling in aortic smooth muscle cells at multiple levels. A potential role for serine phosphorylation in insulin/angiotensin II crosstalk.
Folli F et al.
The Journal of Clinical Investigation, 100, 2158-2158 (1997)
Susanna Pecchia et al.
Plants (Basel, Switzerland), 8(7) (2019-07-25)
Lupins anthracnose is a destructive seed and airborne disease caused by Colletotrichum lupini, affecting stems and pods. Primary seed infections as low as 0.01-0.1% can cause very severe yield losses. One of the most effective management strategies is the development
Manna Crespan et al.
Scientific reports, 10(1), 7206-7206 (2020-05-01)
'Glera' and 'Ribolla Gialla' are the most economically relevant local grapevine cultivars of Friuli Venezia Giulia region (north-eastern Italy). 'Glera' is used to produce the world-renowned Prosecco wine. 'Ribolla Gialla' cultivation is constantly increasing due to the strong demand for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service