81420
Polyvinylpyrrolidone
K 30
Synonym(s):
‘Plasdone’, PVP, Polyvidone, Povidone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
form
powder
application(s)
coatings
pharmaceutical
SMILES string
C=CN1CCCC1=O (Canonical-SMILES)
C=CN1CCCC1=O
InChI
1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2
InChI key
WHNWPMSKXPGLAX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Polyvinylpyrrolidone (PVP), also commercially known as K30, is a water soluble polymer. It has a hygroscopic nature with good adhesive properties. It has a stable pH and has the ability to form transparent films.
Application
PVP has a wide range of usage such as:
- an adhesive for making gluesticks and metal adhesives
- a dispersant for ceramics
- coatings and inks
- formation of synthetic fibres and textiles
- porous membranes
Other Notes
Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Solid dispersion of carbamazepine in PVP K30 by conventional solvent evaporation and supercritical methods
Sethia S and Squillante E
International Journal of Pharmaceutics, 272(1-2), 1-10 (2004)
Robust Ag nanoplate ink for flexible electronics packaging
Li R, et al.
Nanoscale, 7(16), 7368-7377 (2015)
Preparation, characterization and performance studies of polysulfone membranes using PVP as an additive
Chakrabarty B, et al.
Journal of Membrane Science , 315(1-2), 36-47 (2008)
Pressure-dependent conduction behavior of various particles for conductive adhesive applications
Sancaktar E and Dilsiz N
Journal of Adhesion Science and Technology, 13(6), 679-693 (1999)
Photocatalytic microreactor study using TiO 2-coated porous ceramics
Teekateerawej S, et al.
J. Appl. Electrochem., 35(7-8), 693-697 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service