Skip to Content
Merck
All Photos(1)

Key Documents

81430

Sigma-Aldrich

Polyvinylpyrrolidone solution

K 60, 45% in H2O

Synonym(s):

Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solution

concentration

45% in H2O

SMILES string

N1(CCCC1=O)C=C

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Application

Polyvinylpyrrolidone solution (PVP) is also known as K60 and can be used in a variety of applications such as biomedical, tissue engineering, and medical materials.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A pictorial representation of endogenous brain ATP by a bioluminescent method
Kogure K and Alonso OF
Brain Research, 154(2), 273-284 (1978)
Zebrafish embryo xenograft and metastasis assay
Paatero I, et al.
Bio-protocol, 8(18) (2018)
How polysulfone dialysis membranes containing polyvinylpyrrolidone achieve excellent biocompatibility?
Hayama M, et al.
Journal of Membrane Science , 234(1-2), 41-49 (2004)
Chun-Mei Zhao et al.
Environmental toxicology and chemistry, 32(4), 913-919 (2013-01-25)
The toxicity of manufactured silver nanoparticles (AgNPs) has been widely studied, but the influence of AgNPs on the major ions (such as sodium [Na] and calcium [Ca]) regulations are unknown. In the present study, a freshwater cladoceran Daphnia magna was
Liandong Hu et al.
Pakistan journal of pharmaceutical sciences, 26(3), 629-636 (2013-04-30)
Solid dispersion technique has been developed many years for improving solubility of water-insoluble drugs, aiming to achieve a better oral bioavailability. However, this technique exhibits many inconveniences when used for large-scale tableting procedures. The objective of current research work was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service