Skip to Content
Merck
All Photos(4)

Key Documents

N6160

Sigma-Aldrich

Novobiocin sodium

meets USP testing specifications

Synonym(s):

Novobiocin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35N2NaO11
CAS Number:
Molecular Weight:
634.61
Beilstein:
3892910
EC Number:
UNSPSC Code:
51286504
PubChem Substance ID:
NACRES:
NA.76

Agency

USP/NF
meets USP testing specifications

Quality Level

form

powder

antibiotic activity spectrum

Gram-positive bacteria

application(s)

pharmaceutical (small molecule)

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

[Na+].CO[C@@H]1[C@@H](OC(N)=O)[C@@H](O)[C@H](Oc2ccc3C([O-])=C(NC(=O)c4ccc(O)c(C\C=C(\C)C)c4)C(=O)Oc3c2C)OC1(C)C

InChI

1S/C31H35N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34,36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q-1;+1

InChI key

AXOUUAINTJNFRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: coumarin-glycoside

Application

For the production of positively supercoiled plasmid DNA. Inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase. Inhibitor of retrovirus RNA-dependent DNA-polymerase.

Biochem/physiol Actions

Mode of Action: Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II.
Antimicrobial spectrum: Gram-positive bacterial.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiacheng Ma et al.
The Journal of pharmacology and experimental therapeutics, 348(2), 281-292 (2013-11-23)
Impaired neuronal mitochondrial bioenergetics contributes to the pathophysiologic progression of diabetic peripheral neuropathy (DPN) and may be a focal point for disease management. We have demonstrated that modulating heat shock protein (Hsp) 90 and Hsp70 with the small-molecule drug KU-32
Lorenzo González-Molleda et al.
Antimicrobial agents and chemotherapy, 56(2), 893-902 (2011-11-23)
The lytic DNA replication of Kaposi's sarcoma-associated herpesvirus (KSHV) initiates at an origin (ori-Lyt) and requires trans-acting elements, both viral and cellular. We recently demonstrated that several host cellular proteins, including topoisomerases I and II (Topo I and II), are
Jeffery D Eskew et al.
BMC cancer, 11, 468-468 (2011-11-02)
The molecular chaperone, heat shock protein 90 (Hsp90) has been shown to be overexpressed in a number of cancers, including prostate cancer, making it an important target for drug discovery. Unfortunately, results with N-terminal inhibitors from initial clinical trials have
Raphael J Gübeli et al.
Acta biomaterialia, 9(9), 8272-8278 (2013-05-21)
Biohybrid materials combining synthetic polymers with biological components are highly suited for tissue engineering in order to emulate the behavior of natural materials such as the extracellular matrix (ECM). In order to allow for an optimal cell-material interplay, the physical
Huiping Zhao et al.
Bioorganic & medicinal chemistry letters, 23(2), 552-557 (2012-12-14)
Hsp90 is a promising therapeutic target for the treatment of cancer. Novobiocin is the first Hsp90 C-terminal inhibitor ever identified and recent structure-activity relationship studies on the noviose sugar identified several commercially available amines as suitable surrogates. In an effort

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service