Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Novobiocin sodium
Synonym(s):
Novobiocin sodium salt
Empirical Formula (Hill Notation):
C
31
H
35
N
2
NaO
11
CAS Number:
1476-53-5
Molecular Weight:
634.61
EC Number:
216-023-6
Products
Product Category
antibiotics (1)
Brand
Sigma-Aldrich (1)
Feature
Formula Weight
Physical Form
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
N6160
Novobiocin sodium,
meets USP testing specifications
Expand
View Pricing