Skip to Content
Merck
All Photos(1)

Key Documents

46531

Supelco

Novobiocin sodium salt

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35N2NaO11
CAS Number:
Molecular Weight:
634.61
Beilstein:
3892910
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

Assay

97.9% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-positive bacteria

application(s)

clinical testing

format

neat

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

[Na+].CO[C@@H]1[C@@H](OC(N)=O)[C@@H](O)[C@H](Oc2ccc3C([O-])=C(NC(=O)c4ccc(O)c(C\C=C(\C)C)c4)C(=O)Oc3c2C)OC1(C)C

InChI

1S/C31H35N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34,36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q-1;+1

InChI key

AXOUUAINTJNFRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: coumarin-glycoside

Application

For the production of positively supercoiled plasmid DNA. Inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase. Inhibitor of retrovirus RNA-dependent DNA-polymerase.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Mode of Action: Inhibits DNA synthesis by inhibiting the enzyme Topoisomerase II.
Antimicrobial spectrum: Gram-positive bacterial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael J Gübeli et al.
Acta biomaterialia, 9(9), 8272-8278 (2013-05-21)
Biohybrid materials combining synthetic polymers with biological components are highly suited for tissue engineering in order to emulate the behavior of natural materials such as the extracellular matrix (ECM). In order to allow for an optimal cell-material interplay, the physical
Lorenzo González-Molleda et al.
Antimicrobial agents and chemotherapy, 56(2), 893-902 (2011-11-23)
The lytic DNA replication of Kaposi's sarcoma-associated herpesvirus (KSHV) initiates at an origin (ori-Lyt) and requires trans-acting elements, both viral and cellular. We recently demonstrated that several host cellular proteins, including topoisomerases I and II (Topo I and II), are
Jiacheng Ma et al.
The Journal of pharmacology and experimental therapeutics, 348(2), 281-292 (2013-11-23)
Impaired neuronal mitochondrial bioenergetics contributes to the pathophysiologic progression of diabetic peripheral neuropathy (DPN) and may be a focal point for disease management. We have demonstrated that modulating heat shock protein (Hsp) 90 and Hsp70 with the small-molecule drug KU-32
Jeffery D Eskew et al.
BMC cancer, 11, 468-468 (2011-11-02)
The molecular chaperone, heat shock protein 90 (Hsp90) has been shown to be overexpressed in a number of cancers, including prostate cancer, making it an important target for drug discovery. Unfortunately, results with N-terminal inhibitors from initial clinical trials have
David Patrick Kateete et al.
PloS one, 7(9), e45741-e45741 (2012-10-03)
Rhomboids are ubiquitous proteins with unknown roles in mycobacteria. However, bioinformatics suggested putative roles in DNA replication pathways and metabolite transport. Here, mycobacterial rhomboid-encoding genes were characterized; first, using the Providencia stuartii null-rhomboid mutant and then deleted from Mycobacterium smegmatis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service