Skip to Content
Merck
All Photos(1)

Key Documents

ERMAC060

Aflatoxin G2 solution

3.80 μg/g in acetonitrile, ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Beilstein:
1692017
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

concentration

3.80 μg/g in acetonitrile

application(s)

general analytical

format

single component solution

storage temp.

2-8°C

SMILES string

COc1cc2O[C@H]3OCC[C@H]3c2c4OC(=O)C5=C(CCOC5=O)c14

InChI

1S/C17H14O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WPCVRWVBBXIRMA-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMAC060

Legal Information

ERM is a registered trademark of European Commission

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viresh Mohanlall et al.
Journal of food protection, 69(9), 2224-2229 (2006-09-26)
Phytoalexins (stress-induced compounds) from Citrus sinensis cultivar Valencia were screened for antifungal and antimycotoxic activity against a test organism (Cladosporium cladosporoides) and mycotoxin-producing fungi Fusarium verticillioides and Aspergillus parasiticus. The active compound, a member of the coumarin family of compounds
Ludwik Czerwiecki et al.
Roczniki Panstwowego Zakladu Higieny, 58(3), 489-501 (2008-02-06)
The method for determination of aflatoxin B1, B2, G1 i G2 in nuts, culinary spices, cereals and cereal products was described. To optimize the analytical procedure in several products, condition of proper extraction, clean-up, HPLC and detection were selected. After
Meri Kokkonen et al.
Food additives and contaminants, 22(5), 449-456 (2005-07-16)
A simple and feasible method is described for analysing nine mycotoxins in cheese matrix. The method involves liquid extraction followed by high performance liquid chromatographic separation and mass spectrometric detection of the analytes, and allows the determination of aflatoxins B1
Jinyang Zhang et al.
Hybridoma (2005), 28(1), 67-70 (2009-02-26)
Using semisolid methylcellulose HAT medium selection and non-competitive and competitive indirect enzyme-linked immunosorbent assay, three hybridomas that secreted antibodies for aflatoxin G(1) were selected after fusion of mouse SP2/0 myeloma cells with spleen cells isolated from BALB/c mice that had
A A Kalinichenko et al.
Bioorganicheskaia khimiia, 36(1), 122-132 (2010-04-14)
A panel of ten monoclonal antibodies to aflatoxins B1, B2, and G2 was produced and comprehensively characterized. The affinity and cross reactivity of these antibodies were determined using the methods of direct, indirect, and competitive ELISA. The structures of monoclonal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service