Skip to Content
Merck
All Photos(1)

Key Documents

ERMAC058

Aflatoxin B2 solution

3.80 μg/g in acetonitrile, ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O6
CAS Number:
Molecular Weight:
314.29
Beilstein:
1355115
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

concentration

3.80 μg/g in acetonitrile

application(s)

general analytical

format

single component solution

storage temp.

2-8°C

SMILES string

[H][C@@]12CCO[C@]1([H])Oc3cc(OC)c4C5=C(C(=O)CC5)C(=O)Oc4c23

InChI

1S/C17H14O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WWSYXEZEXMQWHT-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMAC058

Legal Information

ERM is a registered trademark of European Commission

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Atehnkeng et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 25(10), 1264-1271 (2008-07-09)
Aflatoxin contamination resulting from maize infection by Aspergillus flavus is both an economic and a public health concern. Therefore, strategies for controlling aflatoxin contamination in maize are being investigated. The abilities of eleven naturally occurring atoxigenic isolates in Nigeria to
J C Netto-Ferreira et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 10(10), 1701-1708 (2011-08-13)
Aflatoxins are mycotoxins produced by fungi of the genus Aspergillus, which is widely spread in the tropics and subtropics. To date, aflatoxin phototoxicity has been recognized, but the mechanism responsible for this phototoxicity has not been fully characterized. In the
D Tang et al.
Biosensors & bioelectronics, 25(2), 514-518 (2009-08-25)
A novel membrane-based lateral-flow immunodipstick assay was developed for the fast screening of aflatoxin B(2) (AFB(2)) as a model compound in food samples. The detector reagent consisted of magnetic nanogold microspheres (MnGMs) with nano-Fe(2)O(3) particles as core and gold nanoparticles
J Yang et al.
Poultry science, 91(11), 2792-2801 (2012-10-24)
The purpose of this study was to evaluate the effects of feeding corn naturally contaminated with aflatoxin B(1) (AFB(1)) and aflatoxin B(2) (AFB(2)) on serum biochemical parameters, hepatic antioxidant enzyme activities, and pathological lesions of broilers. In total, 1,200 Cobb
Debopam Acharya et al.
Analytica chimica acta, 630(1), 82-90 (2008-12-11)
Broad-specific immunoassays are an emerging technology that allows simultaneous determination of a class of closely related small molecular weight analytes. The current approaches are mainly based on the competitive format and involve complicated or expensive processes hindering their wide application.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service