Skip to Content
Merck
All Photos(2)

Key Documents

C7970

Millipore

Casein Peptone

Acid digest of casein from bovine milk, ≥4.6% amino, suitable for biotechnology and microbiology

Synonym(s):

Peptone from casein, Casein acid hydrolysate vitamin free

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

Product Name

Casein acid hydrolysate vitamin free, suitable for microbiology, rich in amino acids and serves as a nitrogen source.

biological source

bovine milk

Quality Level

form

powder

nitrogen analysis

≥4.6% amino
≥7% total

ash

≤42%

loss

≤5% loss on drying

color

off-white to yellow

pH

5-7 (2% in H2O)

application(s)

food and beverages
veterinary

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Casein Acid Hydrolysate is obtained by degrading casein with hydrochloric acid; vitamins and other growth-promoting compounds are largely destroyed in the process. However, it is a rich source of amino acids and serves as a nitrogen source. It is suitable for the preparation of culture media employed in the bacteriological assay of vitamins and antibiotic sensitivity.

Application

Casein Acid Hydrolysate that is free of vitamins can be used as a nitrogen source in culture media that is being used to conduct a bacteriological assay on vitamins and antibiotic sensitivity.

Other Notes

A hydrochloric acid digest of casein free of vitamins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service