Skip to Content
Merck
All Photos(1)

Documents

70166

Millipore

Skim Milk Powder

suitable for microbiology

Synonym(s):

Milk powder fat-free

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

biological source

bovine milk

Quality Level

sterility

non-sterile

form

powder

composition

total nitrogen (N), 4.7-6.0%

availability

not available in USA and Canada

impurities

≤1.5% lipid
≥50.0% reducing sugars (as lactose monohydrate)

ign. residue

≤10%

loss

≤5% loss on drying

application(s)

food and beverages
pathogen testing

microbiology

General description

Skim Milk powder is used for the demonstration of coagulation and proteolysis of casein. Skim Milk is sometimes used as a complete medium or as an ingredient in other media used for the propagation of organisms occurring in milk products. The addition of skim milk to any nutrient-rich medium creates favorable conditions for the growth of organisms, which are encountered in milk. The number of bacteria isolated thus is more than the number of organisms isolated on a regular medium. Proteolytic bacteria hydrolyze casein to form soluble nitrogenous compounds indicated as a clear zone surrounding the colonies on the agar medium. Skim milk serves as a good source of casein

Application

Skim Milk Powder is recommended for cultivation and enumeration of microorganisms encountered in dairy industry

Reconstitution

Mix the powder to a smooth paste with a small quantity of distilled water, then gradually add more distilled water until a 10% w/v mixture is obtained. This is equivalent to fresh milk, and may be sterilized by autoclaving for 5 minutes at 121 °C. Care should be taken not to overheat during sterilization, otherwise caramelization will occur.

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sevil Arabaci Tamer et al.
Peptides, 107, 1-9 (2018-07-22)
Testicular torsion causes ischemia-reperfusion injury and an increased risk of infertility. Nesfatin-1 is a novel peptide with antioxidant, anti-inflammatory and anti-apoptotic properties. In the present study, we aimed to investigate the putative beneficial effects of nesfatin-1 on oxidative injury and
Yun-Hsiang Chen et al.
Scientific reports, 7, 46301-46301 (2017-04-08)
Methamphetamine (Meth) is one of the most frequently abused drugs worldwide. Recent studies have indicated that antibodies with high affinity for Meth reduce its pharmacological effects. The purpose of this study was to develop a technique for virus-based passive immunization
Jikai Liu et al.
International journal of oncology, 50(2), 356-364 (2016-12-22)
miR‑223‑3p is deregulated in several tumor types and plays an important role in tumorigenesis and progression. However, its role in the pathogenesis of testicular germ cell tumor (TGCT) remains uncharacterized. We previously demonstrated that miR‑223‑3p expression was increased in TGCTs
Fabian Sesterhenn et al.
PLoS biology, 17(2), e3000164-e3000164 (2019-02-23)
Throughout the last several decades, vaccination has been key to prevent and eradicate infectious diseases. However, many pathogens (e.g., respiratory syncytial virus [RSV], influenza, dengue, and others) have resisted vaccine development efforts, largely because of the failure to induce potent
Madalina E Carter-Timofte et al.
Genes and immunity, 20(3), 214-223 (2018-05-08)
Recently, deficiency in the cytosolic DNA sensor RNA Polymerase III was described in children with severe primary varicella-zoster virus (VZV) infection in the CNS and lungs. In the present study we examined adult patients with VZV CNS infection caused by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service