Skip to Content
Merck
All Photos(2)

Key Documents

51841

Millipore

Peptone (vegetable) acid hydrolysate

suitable for microbiology

Synonym(s):

Peptone from vegetable, Casein acid hydrolysate replacement

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

biological source

plant

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥3%
total nitrogen (N), ≥6%

impurities

≤44% ash

loss

≤5% loss on drying

pH

5.5±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, yellow

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Peptone (vegetable) acid hydrolysate is a purified hydrolysate which provides the necessary nitrogenous material for culture media or can be used when amino acid mixtures are specified. It can be used for preparing Antibiotic Sensitivity Test Media including Mueller Hinton Agar, in media requiring quantitative addition of tryptophan, in vaccine preparation media as a source of high concentration of free amino acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naila Emanuil et al.
Plants (Basel, Switzerland), 9(12) (2020-12-24)
The accumulation of cadmium (Cd) in edible plant parts and fertile lands is a worldwide problem. It negatively influences the growth and productivity of leafy vegetables (e.g., spinach, Spinacia oleracea L.), which have a high tendency to radially accumulate Cd.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service