Skip to Content
Merck
All Photos(1)

Key Documents

C2160000

Chymotrypsin

BRP, European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chymotrypsin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M V Kolodzeĭskaia et al.
Ukrainskii biokhimicheskii zhurnal (1978), 60(4), 103-117 (1988-07-01)
Recent data on the nature of trypsin-, chymotrypsin-like proteinases of fish are generalized. Localization and secretion of these enzymes in pyloric appendages of fish are considered in detail. Trypsin and chymotrypsin are in the state of proenzymes and transform into
[The use of proteolytic enzymes (trypsin, chymotrypsin, streptokinase, streptodornase, papain) in stomatology].
A I Protasevich
Stomatologiia, 46(5), 107-109 (1967-09-01)
THE CHEMICAL MODIFICATION OF CHYMOTRYPSIN.
G H DIXON et al.
Canadian journal of biochemistry, 42, 695-714 (1964-05-01)
[Stages of hydrolysis catalyzed by trypsin and chymotrypsin].
V M Krutiakov
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, 5, 671-680 (1973-09-01)
Evolutionary divergence of substrate specificity within the chymotrypsin-like serine protease fold.
J J Perona et al.
The Journal of biological chemistry, 272(48), 29987-29990 (1997-12-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service