Skip to Content
Merck
All Photos(5)

Documents

C4129

Sigma-Aldrich

α-Chymotrypsin from bovine pancreas

Type II, lyophilized powder, ≥40 units/mg protein

Synonym(s):

α-chymotrypsin A and B, alpha-chymotrypsin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010112
NACRES:
NA.54

type

Type II

form

lyophilized powder

specific activity

≥40 units/mg protein

mol wt

25 kDa

composition

protein, ≥85%

color

white to off-white

UniProt accession no.

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The enzyme from Sigma has been used to study the structure-function relationship in glycosylated α-chymotrypsin using immobilized metal-ion affinity chromatography (IMAC) and immobilized metal-ion affinity capillary electrophoresis (IMACE).

Biochem/physiol Actions

α-Chymotrypsin is a serine peptidase and has 241 amino acid residues contained in three polypeptide chains (A chain-13 residues, B chain-131 residues, and C chain-97 residues) linked by disulfide bridges. Molecular weight of this enzyme is found to be 25 kDa. The pI is 8.75. It selectively hydrolyzes peptide bonds on the C-terminal side of tyrosine, phenylalanine, tryptophan, and leucine. Ca2+ activates and stabilizes the enzyme. The enzyme is inhibited by diisopropyl fluorophosphate (DFP), phenylmethanesulfonyl fluoride (PMSF), N-p-tosyl-L-phenylalanine chloromethyl ketone (TPCK), chymostatin, aprotinin, α1-antitrypsin, and α2-macroglobulin, 10 mM Cu2+ and Hg2+.
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met, Leu) on the carboxyl end of the bond.

Unit Definition

One unit will hydrolyze 1.0 μmole of BTEE per min at pH 7.8 at 25 °C.

Preparation Note

Produced from 3× crystallized chymotrypsinogen

Analysis Note

Protein determined by E1%/280

Other Notes

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ines Atuh Ngoh et al.
Experimental parasitology, 218, 107969-107969 (2020-08-29)
Invasion of human red blood cells (RBCs) by Plasmodium parasites is a crucial yet poorly characterised phenotype. Two-color flow cytometry (2cFCM) promises to be a very sensitive and high throughput method for phenotyping parasite invasion. However, current protocols require high
Xingchen Chen et al.
Marine drugs, 17(12) (2019-12-18)
Serine proteases play pivotal roles in normal physiology and a spectrum of patho-physiological processes. Accordingly, there is considerable interest in the discovery and design of potent serine protease inhibitors for therapeutic applications. This led to concerted efforts to discover versatile
Olivia Ogilvie et al.
MethodsX, 7, 101076-101076 (2020-10-10)
Mass spectrometry (MS) is an emerging method to determine the accurate concentration of immunogenic gluten peptides. It is of interest to quantify specific peptides within the gluten peptidome due to the role they play in the activation of the celiac
Antonio Jesús Vizcaíno et al.
Marine drugs, 18(6) (2020-06-24)
This piece of research evaluates the presence of protease inhibitors in the macroalga Ulva ohnoi and provides an initial overview of their mode of action. The ability of Ulva protease inhibitors to inhibit digestive proteases of three marine fish species
Shannon Gwala et al.
Food & function, 11(3), 2509-2521 (2020-03-07)
Postharvest storage of legumes is a strategy to ensure food security. However, the hard-to-cook phenomenon, partly explained by pectin-cation-phytate theory, may develop during storage. Based on this theory, minerals could be redistributed within the matrix, affecting the concentration of free/bound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service