Skip to Content
Merck

Skip To

C3142

α-Chymotrypsin from bovine pancreas

(TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein

Synonym(s):

TLCK-Chymotrypsin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
42010112
EC Number:
232-671-2
NACRES:
NA.54
MDL number:
EC Number:
Specific activity:
≥40 units/mg protein
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

type

Type VII

form

essentially salt-free, lyophilized powder

specific activity

≥40 units/mg protein

mol wt

25 kDa

solubility

1 mM HCl: soluble 10 mg/mL, clear

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... CTRB1(618826)

Application

α-Chymotrypsin from Sigma has been used to determine the crystal structures of two homologous inhibitors (pars intercerebralis major peptide-C and pars intercerebralis major peptide-D2v) from the insect Locusta migratoria by forming a complex with the enzyme.

Biochem/physiol Actions

α-Chymotrypsin is a serine peptidase and has 241 amino acid residues contained in three polypeptide chains (A chain-13 residues, B chain-131 residues, and C chain-97 residues) linked by disulfide bridges. Molecular weight of this enzyme is found to be 25 kDa. Its pI is 8.75. It selectively hydrolyzes peptide bonds on the C-terminal side of tyrosine, phenylalanine, tryptophan, and leucine. Ca2+ activates and stabilizes the enzyme. The enzyme is inhibited by diisopropyl fluorophosphate (DFP), phenylmethanesulfonyl fluoride (PMSF), N-p-tosyl-L-phenylalanine chloromethyl ketone (TPCK), chymostatin, aprotinin, α1-antitrypsin, α2-macroglobulin, 10 mM Cu2+ and Hg2+.
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met, Leu) on the carboxyl end of the bond.

Preparation Note

TLCK treatment inactivates trypsin which may be present in chymotrypsin, without affecting the chymotrypsin activity.

Analysis Note

Protein determined by A1%/280

Other Notes

One unit will hydrolyze 1.0 μmole of BTEE per min at pH 7.8 at 25 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7762C4129CHY5S
description

(TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein

description

Type I-S, essentially salt-free, lyophilized powder

description

Type II, lyophilized powder, ≥40 units/mg protein

description

≥40 units/mg protein, vial of 5 mg

specific activity

≥40 units/mg protein

specific activity

≥40 units/mg protein

specific activity

≥40 units/mg protein

specific activity

≥40 units/mg protein

Gene Information

cow ... CTRB1(618826)

Gene Information

cow ... CTRB1(618826)

Gene Information

cow ... CTRB1(618826)

Gene Information

cow ... CTRB1(618826)

form

essentially salt-free, lyophilized powder

form

essentially salt-free, lyophilized powder

form

lyophilized powder

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

UniProt accession no.

P00767

UniProt accession no.

P00767

UniProt accession no.

P00767

UniProt accession no.

P00767

solubility

1 mM HCl: soluble 10 mg/mL, clear

solubility

1 mM HCl: soluble 2.0 mg/mL, clear

solubility

-

solubility

-


Still not finding the right product?


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 1

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

1–5 of 5 Questions  
  1. What is the molecular weight of alpha-chymotrypsin from bovine pancreas (Product No. C3142)?

    1 answer
    1. The molecular weight of alpha chymotrypsin, reported in the literature, is approximately 25 kDa.This information and the basic structure description of the enzyme can be found on our product information sheet (under Documents, above) at our website.

      Helpful?

  2. What is the pH optimum for the enzyme, Product No. C3142?

    1 answer
    1. The pH optimum for alpha-chymotrypsin is between pH 7.5-8.5. Please see the following reference: Wilcox, P.E., Chymotrypsingogens-chymotrypsins, Methods in Enzymology, 19, 64-80 (1970).

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How can solutions of alpha-chymotrypsin (Product No. C3142) be stored?

    1 answer
    1. Stock solutions prepared in 1 mM HCl, containing 2 mM calcium chloride, can be stored at -20 ° for about one week.

      Helpful?

  5. How do you recommend preparing a solution of Product No. C3142?

    1 answer
    1. It is recommended to reconstitute Product No. C3142 in 1 mM hydrochloric acid containing 2 mM calcium chloride. The calcium functions as both a stabilizer (and possibly an activator) of the enzyme.

      Helpful?

Reviews

No rating value

Active Filters