Skip to Content
Merck
All Photos(1)

Documents

A1202000

Citric acid

anhydrous, European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP reference standard

API family

citric acid

quality

anhydrous

expl. lim.

8 %, 65 °F

manufacturer/tradename

EDQM

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Citric acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levente Karaffa et al.
Applied microbiology and biotechnology, 61(3), 189-196 (2003-04-17)
This Mini-Review summarizes the current knowledge on the biochemical and physiological events leading to massive citric acid accumulation by Aspergillus niger under industrially comparable conditions, thereby particularly emphasizing the roles of glycolytic flux and its control, excretion of citric acid
Yohei Nakayama et al.
Journal of oral science, 56(4), 261-268 (2014-12-17)
Amelotin (AMTN) is a secreted protein transcribed predominantly during the maturation stage of enamel formation and localized in the junctional epithelium. We investigated differences in the levels of AMTN gene expression between non-inflamed gingiva and inflamed gingiva from patients with
Wei-An Lai et al.
Journal of molecular endocrinology, 53(2), 259-270 (2014-07-25)
Estrogens are essential for female reproduction and overall well-being, and estrogens in the circulation are largely synthesized in ovarian granulosa cells. Using primary cultures of ovarian granulosa cells from gonadotropin-primed immature rats, we have recently discovered that pituitary FSH and
Giampiero Rossi-Fedele et al.
Journal of endodontics, 38(4), 426-431 (2012-03-15)
Root canal irrigants play a significant role in the elimination of microorganisms, tissue dissolution, and the removal of debris and smear layer. No single solution is able to fulfill these actions completely; therefore, their association is required. The aim of
Konrad Hoetzenecker et al.
European heart journal, 36(11), 676-685 (2013-01-17)
Supernatants of serum-free cultured mononuclear cells (MNC) contain a mix of immunomodulating factors (secretome), which have been shown to attenuate detrimental inflammatory responses following myocardial ischaemia. Inflammatory dilated cardiomyopathy (iDCM) is a common cause of heart failure in young patients.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service