Merck
  • Antagonistic interactions between sodium hypochlorite, chlorhexidine, EDTA, and citric acid.
Antagonistic interactions between sodium hypochlorite, chlorhexidine, EDTA, and citric acid.
Journal of endodontics (2012-03-15)
Giampiero Rossi-Fedele, Esma J Doğramaci, Andrea R Guastalli, Liviu Steier, Jose Antonio Poli de Figueiredo
ABSTRACT
Root canal irrigants play a significant role in the elimination of microorganisms, tissue dissolution, and the removal of debris and smear layer. No single solution is able to fulfill these actions completely; therefore, their association is required. The aim of this investigation was to review the antagonistic interactions occurring when sodium hypochlorite (NaOCl), chlorhexidine (CHX), EDTA, and citric acid (CA) are used together during endodontic treatment. A search was performed in the electronic database Medline (articles published through 2011; English language; and the following search terms or combinations: "interaction AND root canal irrigant or endodontic irrigant or sodium hypochlorite or chlorhexidine," "sodium hypochlorite AND EDTA or ethylenediaminetetraacetic acid or citric acid or chelating agent or chlorhexidine," and "chlorhexidine AND EDTA or ethylenediaminetetraacetic acid or citric acid or chelating agent") to identify publications that studied unwanted chemical interactions between NaOCl, CHX, and EDTA and CA. The search identified 1,285 publications; 19 fulfilled the inclusion/exclusion criteria of the review. Their research methodology was classified as either in vitro or ex vivo. Antagonistic interactions included the loss of free available chlorine for NaOCl when in contact with chelators, which consequently reduced the tissue dissolution capability and to a lesser extent antimicrobial activities. When CHX and NaOCl are mixed, a precipitate forms that can present detrimental consequences for endodontic treatment, including a risk of discoloration and potential leaching of unidentified chemicals into the periradicular tissues. CHX and EDTA mixtures cause a precipitate, whereas CHX and CA do not exhibit interaction.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%