Merck
  • Calcineurin and CRTC2 mediate FSH and TGFβ1 upregulation of Cyp19a1 and Nr5a in ovary granulosa cells.
Calcineurin and CRTC2 mediate FSH and TGFβ1 upregulation of Cyp19a1 and Nr5a in ovary granulosa cells.
Journal of molecular endocrinology (2014-07-25)
Wei-An Lai, Yi-Ting Yeh, Wei-Ling Fang, Leang-Shin Wu, Nobuhiro Harada, Peng-Hui Wang, Ferng-Chun Ke, Wen-Ling Lee, Jiuan-Jiuan Hwang
ABSTRACT
Estrogens are essential for female reproduction and overall well-being, and estrogens in the circulation are largely synthesized in ovarian granulosa cells. Using primary cultures of ovarian granulosa cells from gonadotropin-primed immature rats, we have recently discovered that pituitary FSH and ovarian cytokine transforming growth factor beta 1 (TGFβ1) induce calcineurin-mediated dephosphorylation-activation of cAMP-response element-binding protein (CREB)-regulated transcription coactivator (CRTC2) to modulate the expression of Star, Cyp11a1, and Hsd3b leading to increased production of progesterone. This study explored the role of calcineurin and CRTC2 in FSH and TGFβ1 regulation of Cyp19a1 expression in granulosa cells. Ovarian granulosa cells treated with FSH displayed increased aromatase protein at 24  h post-treatment, which subsided by 48  h, while TGFβ1 acting through its type 1 receptor augmented the action of FSH with a greater and longer effects. It is known that the ovary-specific Cyp19a1 PII-promoter contains crucial response elements for CREB and nuclear receptor NR5A subfamily liver receptor homolog 1 (LRH1/NR5A2) and steroidogenic factor 1 (SF1/NR5A1), and that the Nr5a2 promoter also has a potential CREB-binding site. Herein, we demonstrate that FSH+TGFβ1 increased LRH1 and SF1 protein levels, and their binding to the Cyp19a1 PII-promoter evidenced, determined by chromatin immunoprecipitation analysis. Moreover, pretreatment with calcineurin auto-inhibitory peptide (CNI) abolished the FSH+TGFβ1-upregulated but not FSH-upregulated aromatase activity at 48  h, and the corresponding mRNA changes in Cyp19a1, and Nr5a2 and Nr5a1 at 24  h. In addition, FSH and TGFβ1 increased CRTC2 binding to the Cyp19a1 PII-promoter and Nr5a2 promoter at 24  h, with CREB bound constitutively. In summary, the results of this study indicate that calcineurin and CRTC2 have important roles in mediating FSH and TGFβ1 collateral upregulation of Cyp19a1 expression together with its transcription regulators Nr5a2 and Nr5a1 in ovarian granulosa cells.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%