Merck
  • Aspergillus niger citric acid accumulation: do we understand this well working black box?
Aspergillus niger citric acid accumulation: do we understand this well working black box?
Applied microbiology and biotechnology (2003-04-17)
Levente Karaffa, Christian P Kubicek
ABSTRACT
This Mini-Review summarizes the current knowledge on the biochemical and physiological events leading to massive citric acid accumulation by Aspergillus niger under industrially comparable conditions, thereby particularly emphasizing the roles of glycolytic flux and its control, excretion of citric acid from the mitochondria and the cytosol, and the critical fermentation variables. The potential of novel techniques for metabolic analysis and genomic approaches in understanding this fermentation is also discussed.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%