Merck
  • The voltage-dependent gate in MthK potassium channels is located at the selectivity filter.
The voltage-dependent gate in MthK potassium channels is located at the selectivity filter.
Nature structural & molecular biology (2012-12-25)
David J Posson, Jason G McCoy, Crina M Nimigean
ABSTRACT
Understanding how ion channels open and close their pores is crucial for comprehending their physiological roles. We used intracellular quaternary ammonium blockers, electrophysiology and X-ray crystallography to locate the voltage-dependent gate in MthK potassium channels from Methanobacterium thermoautotrophicum. Blockers bind in an aqueous cavity between two putative gates: an intracellular gate and the selectivity filter. Thus, these blockers directly probe gate location--an intracellular gate will prevent binding when closed, whereas a selectivity filter gate will always allow binding. Kinetic analysis of tetrabutylammonium block of single MthK channels combined with X-ray crystallographic analysis of the pore with tetrabutyl antimony unequivocally determined that the voltage-dependent gate, like the C-type inactivation gate in eukaryotic channels, is located at the selectivity filter. State-dependent binding kinetics suggest that MthK inactivation leads to conformational changes within the cavity and intracellular pore entrance.
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%