Skip to Content
Merck
All Photos(2)

Documents

95054

Supelco

Azelaic acid

analytical standard

Synonym(s):

Nonanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)7CO2H
CAS Number:
Molecular Weight:
188.22
Beilstein:
1101094
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

6.5 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

bp

286 °C/100 mmHg (lit.)

mp

106-112 °C
109-111 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(=O)CCCCCCCC(O)=O

InChI

1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)

InChI key

BDJRBEYXGGNYIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azelaic acid (heptane-l,7-dicarboxylic acid), a dicarboxylic saturated acid, is an active component of some topical anti-acne preparations. It is reportedly identified in cosmetics and pharmaceuticals. Azelaic acid is found to suppress the growth of Propionibacterium spp. and reduce keratinization.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Azelaic acid may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Tobacco using gas chromatography coupled to mass spectrometry (GC-MS).
  • Topical preparations using reversed-phase liquid chromatographic (LC) with UV-Vis detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of azelaic and benzoic acids in topical preparations by liquid chromatography
Mansour MA and Ibrahiem MM
Chromatographia, 55(7-8), 435-437 (2002)
Simultaneous determination of non?volatile, semi?volatile, and volatile organic acids in tobacco by SIM?Scan mode GC?MS
Wang B, et al.
Journal of Separation Science, 31(4), 721-726 (2008)
Syntheses and QSAR studies of sorbic, cinnamic and ricinoleic acid derivatives as potential antibacterial agents
Narasimhan B, et al.
Indian Journal of Chemistry, 42B, 2828-2834 (2003)
Dongdong Zhang et al.
Oxidative medicine and cellular longevity, 2020, 1295984-1295984 (2021-01-12)
Acute myeloid leukemia (AML) is a hematological malignancy with a poor prognosis attributed to elevated reactive oxygen species (ROS) levels. Thus, agents that inhibit ROS generation in AML should be exploited. Azelaic acid (AZA), a small molecular compound, can scavenge
Aditya K Gupta et al.
International journal of dermatology, 46(5), 533-538 (2007-05-03)
In December of 2002, the FDA approved azelaic acid 15% gel for the topical treatment of inflammatory papules and pustules of mild to moderate rosacea. Azelaic acid is a saturated dicarboxylic acid, which is naturally occurring and has been used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service