Skip to Content
Merck
All Photos(3)

Documents

O2136

Sigma-Aldrich

Oleamide

≥99% (TLC), powder, sleep-inducing brain lipid

Synonym(s):

cis-9,10-Octadecenoamide, cis-9-Octadecenamide, Oleic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35NO
CAS Number:
Molecular Weight:
281.48
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Oleamide, ≥99%

Assay

≥99%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(N)=O

InChI

1S/C18H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H2,19,20)/b10-9-

InChI key

FATBGEAMYMYZAF-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oleamide is a lipid or a brain fatty acid, that is found in the CSF (cerebrospinal fluid). It is originally obtained from the cerebrospinal fluid of cats, that are sleep-deprived.

Application

Oleamide has been used as a supplement in glucose and galactose media to prevent the rescue of galactose-induced Leigh syndrome (LS).

Biochem/physiol Actions

Oleamide has the ability to stimulate sleep. It possess several properties, like cannabinoid-like activity. Oleamide can also disable the communication, facilitated by gap junction between rat glial cells.
Sleep-inducing brain lipid, which allosterically modulates GABAA receptors and potentiates 5-HT7 serotonin receptor responses. Selective endogenous agonist of rat and human CB1 cannabinoid receptor.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Varadarajan Sudhahar et al.
European journal of pharmacology, 607(1-3), 143-150 (2009-03-28)
Fatty acid amides are a new class of signaling lipids that have been implicated in diverse physiological and pathological conditions. Oleamide is a fatty acid amide that induces vasorelaxation. Here, we investigated the mechanisms behind the vasorelaxation effect of oleamide
Edgar Soria-Gómez et al.
The international journal of neuropsychopharmacology, 13(9), 1247-1254 (2010-07-29)
The central nervous system control of food intake has been extensively studied, hence, several neurotransmitter systems regulating this function are now clearly identified, for example, the endocannabinoid and serotoninergic systems. The former stimulates feeding while the latter inhibits it. Oleamide
Andrea Herrera-Solís et al.
Pharmacology, biochemistry, and behavior, 95(1), 106-112 (2010-01-09)
Anandamide and oleamide, induce sleep when administered acutely, via the CB1 receptor. Their subchronic administration must be tested to demonstrate the absence of tolerance to this effect, and that the sudden withdrawal of these endocannabinoids (eCBs) does not affect sleep
Lidong Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 29(1), 65-74 (2012-12-12)
A series of oleamide derivatives, (C(18)H(34)NO)(2)(CH(2))(n) [n = 2 (1a), 3 (1b), 4 (1c), or 6 (1d); C(18)H(34)NO = oleic amide fragment] and (C(18)H(34)NO)(CH(2))(6)NH(2) (2), have been synthesized and their self-assembly is investigated in ethanol/water media. Self-assembly of 1a and
Mónica Méndez-Díaz et al.
Addiction biology, 17(4), 725-735 (2010-12-25)
Endocannabinoids (eCBs) are mediators of the homeostatic and hedonic systems that modulate food ingestion. Hence, eCBs, by regulating the hedonic system, may be modulating the valence of the emotion associated to food ingestion (positive: pleasant or negative: unpleasant). Our first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service