Skip to Content
Merck
All Photos(2)

Documents

A72405

Sigma-Aldrich

2-Amino-1-phenylethanol

98%

Synonym(s):

α-(Aminomethyl)benzyl alcohol, DL-β-Hydroxyphenethylamine, Phenylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OH)C6H5
CAS Number:
Molecular Weight:
137.18
Beilstein:
971222
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

160 °C/17 mmHg (lit.)

mp

56-58 °C (lit.)

SMILES string

NCC(O)c1ccccc1

InChI

1S/C8H11NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H,6,9H2

InChI key

ULSIYEODSMZIPX-UHFFFAOYSA-N

Gene Information

human ... PNMT(5409)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qian Wu et al.
Journal of medicinal chemistry, 48(23), 7243-7252 (2005-11-11)
The X-ray structure of human phenylethanolamine N-methyltransferase (hPNMT) complexed with its product, S-adenosyl-L-homocysteine (4), and the most potent inhibitor reported to date, SK&F 64139 (7), was used to identify the residues involved in inhibitor binding. Four of these residues, Val53
Christine L Gee et al.
Biochimica et biophysica acta, 1750(1), 82-92 (2005-05-17)
The crystal structure of human phenylethanolamine N-methyltransferase (hPNMT) reveals a disulfide-linked dimer, despite the presence of reducing agent in the crystallisation conditions. By removing the reducing agent, hPNMT crystals grow more rapidly and at lower protein concentrations. However, it was
M Weinstock et al.
Neuropharmacology, 43(6), 999-1005 (2002-11-09)
TV-3326 is a novel cholinesterase inhibitor that produces irreversible brain-selective inhibition of monoamine oxidase (MAO)-A and B and has antidepressant-like activity in rats after chronic oral administration. This study determined whether TV-3326 would cause less potentiation than other irreversible MAO-inhibitors
Panayiotis A Procopiou et al.
Journal of medicinal chemistry, 52(8), 2280-2288 (2009-03-26)
A series of saligenin alkoxyalkylphenylsulfonamide beta(2) adrenoceptor agonists were prepared by reacting a protected saligenin oxazolidinone with alkynyloxyalkyl bromides, followed by Sonogashira reaction, hydrogenation, and deprotection. The meta-substituted primary sulfonamide was more potent than the para- and the ortho-analogues. Primary
Masashi Imanishi et al.
Journal of medicinal chemistry, 51(15), 4804-4822 (2008-07-25)
We designed a series of benzoic acid derivatives containing the biphenyl ether or biphenyl template on the RHS and a phenylethanolaminotetraline (PEAT) skeleton, which was prepared by highly stereoselective synthesis, to generate two structurally different lead compounds ( 10c, 10m)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service