Skip to Content
Merck
All Photos(1)

Key Documents

73982

Supelco

Nonanoic acid

analytical standard

Synonym(s):

Acid C9, Pelargonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COOH
CAS Number:
Molecular Weight:
158.24
Beilstein:
1752351
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

9 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.432 (lit.)
n20/D 1.432

bp

268-269 °C (lit.)

mp

9 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCC(O)=O

InChI

1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)

InChI key

FBUKVWPVBMHYJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nonanoic acid may be used as an internal standard for the quantification of valproic acid and other common anticonvulsant drugs in biological samples using high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock.AG
Fenaroli's Handbook of Flavor Ingredients, (4) (2001)
Preparation and characterization of poly (. beta.-hydroxyalkanoates) obtained from Pseudomonas oleovorans grown with mixtures of 5-phenylvaleric acid and n-alkanoic acids
Kim.B.Y, et al.
Macromolecules, 24(19), 5256-5260 (1991)
HPLC method for the simultaneous analysis of valproic acid and other common anticonvulsant drugs in human plasma or serum
Lucarelli.C, et al.
Chromatographia, 33(1-2), 37-40 (1992)
Ingegärd Anveden et al.
Contact dermatitis, 50(5), 298-303 (2004-06-24)
A multicentre, randomized, double-blind, crossover study was designed to investigate the effects of prednisone on allergic and irritant patch test reactions. 24 subjects with known allergy to nickel were recruited and patch tested with a nickel sulfate dilution series in
G J Venter et al.
Veterinary parasitology, 181(2-4), 365-369 (2011-05-20)
The efficacy of a 15% (w/w) mixture of octanoic, nonanoic and decanoic acids in light mineral oil to repel Culicoides biting midges (Diptera; Ceratopogonidae) was determined in three replicates of a 4 × 4 Latin square design under South African

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service