Skip to Content
Merck
All Photos(1)

Key Documents

900682

Sigma-Aldrich

Water

suitable for mass spectrometry (MS) and UHPLC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
NACRES:
NA.05
grade:
HPLC grade
technique(s):
mass spectrometry (MS): suitable
application(s):
PFAS testing
food and beverages
bp:
100 °C (lit.)

Product Name

Water, for UHPLC, suitable for mass spectrometry (MS)

grade

HPLC grade

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1621
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

reg. compliance

suitable for FDA C-010.02

form

liquid

shelf life

limited shelf life,expiry date on the label

technique(s)

mass spectrometry (MS): suitable

refractive index

n20/D 1.34 (lit.)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

SMILES string

O

application(s)

PFAS testing
food and beverages

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Water is used as a solvent for UHPLC-MS with the following advantages:        
  • Highest solvent purity for low detection limits.       
  • Extremely low MS baseline noise.       
  • Exemplary lot-to-lot reproducibility.       
  • Minimal trace metals.        
  • Microfiltered (0.2 μm).

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service