Skip to Content
Merck
All Photos(3)

Key Documents

861146

Sigma-Aldrich

Erioglaucine disodium salt

coloring dye

Synonym(s):

Acid Blue 9, Alphazurine FG, FD&C;BLUE No. 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H34Na2N2O9S3
CAS Number:
Molecular Weight:
792.85
Colour Index Number:
42090
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

form

powder

technique(s)

titration: suitable

color

purple to dark purple

mp

283 °C (dec.) (lit.)

solubility

water: 1 mg/mL

λmax

406 nm
625 nm (2nd)

ε (extinction coefficient)

≥10000 at 306-310 nm in H2O
≥80000 at 627-631 nm in H2O
≥9500 at 627-631 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CCN(Cc1cccc(c1)S([O-])(=O)=O)c2ccc(cc2)C(=C3\C=C/C(C=C3)=[N+](\CC)Cc4cccc(c4)S([O-])(=O)=O)\c5ccccc5S([O-])(=O)=O

InChI

1S/C37H36N2O9S3.2Na/c1-3-38(25-27-9-7-11-33(23-27)49(40,41)42)31-19-15-29(16-20-31)37(35-13-5-6-14-36(35)51(46,47)48)30-17-21-32(22-18-30)39(4-2)26-28-10-8-12-34(24-28)50(43,44)45;;/h5-24H,3-4,25-26H2,1-2H3,(H2-,40,41,42,43,44,45,46,47,48);;/q;2*+1/p-2

InChI key

SGHZXLIDFTYFHQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Erioglaucine disodium salt has been used as a coloring dye to determine the feeding behaviour in Drosophila. It has been used as a coloring dye for determining the number of egg masses of Meloidogyne incognita.

Biochem/physiol Actions

Erioglaucine is an acid aminotriphenylmethane dye with blue color.

Analysis Note

Dye content determined on each lot

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diogo Sousa-Martins et al.
Retina (Philadelphia, Pa.), 32(7), 1328-1336 (2012-03-28)
To determine whether a natural dye solution based on lutein and zeaxanthin alone or combined with Brilliant Blue stains and facilitates peeling of intraocular membranes in human eyes. In this study of 60 cadaveric eyes, open-sky vitrectomy including posterior hyaloid
Xiang Cai et al.
Langmuir : the ACS journal of surfaces and colloids, 27(12), 7828-7835 (2011-05-19)
A water-soluble brilliant blue/reduced graphene oxide/tetradecyltriphenylphosphonium bromide composite (BB-rGO-TTP) was prepared by using noncovalent brilliant blue-functionalized reduced graphene oxide (BB-rGO) as the tetradecyltriphenylphosphonium bromide (TTP) carrier. Antibacterial performance of this novel composite was investigated for both Gram-positive and Gram-negative bacteria.
Altered lipid metabolism in Drosophila model of Huntington's disease.
Aditi K, et al.
Scientific Reports, 6, 31411-31411 (2016)
No evidence for behavioural adaptations to nematode parasitism by the fly Drosophila putrida.
Debban CL and Dyer KA
Journal of environmental biology / Academy of Environmental Biology, India, 26, 1646-1646 (2013)
Gaëlle Coussot et al.
Analytical and bioanalytical chemistry, 399(3), 1061-1069 (2010-11-09)
The covalent immobilization of synthetic or natural macromolecular compounds containing amino groups onto polystyrene (PS) solid surfaces is of great interest in diagnostic applications. A sensitive assay allowing the determination of reactive end groups is therefore a powerful tool for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service