Skip to Content
Merck
All Photos(3)

Key Documents

E12907

Sigma-Aldrich

Ethyl benzoate

≥99%

Synonym(s):

Benzoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOC2H5
CAS Number:
Molecular Weight:
150.17
Beilstein:
1908172
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.17 (vs air)

vapor pressure

1 mmHg ( 44 °C)

Assay

≥99%

form

liquid

autoignition temp.

914 °F

refractive index

n20/D 1.504 (lit.)

bp

212 °C (lit.)

mp

−34 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1ccccc1

InChI

1S/C9H10O2/c1-2-11-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

MTZQAGJQAFMTAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl benzoate is a general reagent to construct molecules with phenyl pendants such as phenyl bearing pyrazine-boron fluorescent complex. It can be used to prepare Horner′s phosphonate intermediate in the total synthesis of diospongins A and B. It is also a flavoring agent used in food and fragrance industry.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and fluorescence properties of novel pyrazine?boron complexes bearing a ?-iminoketone ligand.
Kubota Y, et al.
Organic Letters, 13(24), 6544-6547 (2011)
Densities and viscosities of binary mixtures of isoamyl acetate, ethyl caproate, ethyl benzoate, isoamyl butyrate, ethyl phenylacetate, and ethyl caprylate with ethanol at T=(288.15, 298.15, 308.15, and 318.15) K.
Sheu Y W and Tu C H
Journal of Chemical and Engineering Data, 51(2), 496-503 (2006)
A concise total synthesis of diospongins A and B.
Sabitha G, et al.
Helvetica Chimica Acta, 91(12), 2235-2239 (2008)
Md Munir Mostafiz et al.
Ecotoxicology and environmental safety, 184, 109653-109653 (2019-09-19)
Plant-derived benzoates are known to have insecticidal and repellent activities, however, these effects have been evaluated thoroughly in only a few groups of insects. In this study, the insecticidal activities of three commercially available benzoates, methyl benzoate (MB), ethyl benzoate
Daniel I Perez et al.
Bioorganic & medicinal chemistry, 17(19), 6914-6925 (2009-09-15)
Thienylhalomethylketones, whose chemical, biological, and pharmaceutical data are here reported, are the first irreversible inhibitors of GSK-3beta described to date. Their inhibitory activity is likely related to the cysteine residue present in the ATP-binding site, which is proposed as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service