Skip to Content
Merck
All Photos(1)

Documents

75832

Millipore

Coagulase Test (Slide)

suitable for microbiology

Synonym(s):

Slide Coagulase Test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

Quality Level

form

disc

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

Staphylococcus spp.

Application

Coagulase Test (Slide) can be used for the detection of coagulase-negative or positive organisms (Staphylococcus aureus).

Packaging

1 box contains 30 discs (made from rabbit plasma)

Principle

Coagulase Test is carried out to differentiate potentially pathogenic Staphylococcus species from other Gram positive, catalase-positive cocci by the detection of coagulase. It is an important indicator for the pathogenicity of Staphylococcus strains.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service