Skip to Content
Merck
All Photos(1)

Key Documents

40560

Millipore

Oxidase Test Strips

suitable for agriculture, clinical testing, environmental, food and beverages, pharmaceutical, microbiology and specific enzyme detection

Synonym(s):

Cytochrome oxidase activity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
eCl@ss:
32161000
NACRES:
NA.85
Pricing and availability is not currently available.

Product Name

Oxidase Strips, suitable for microbiology

Agency

according to ISO 9308-1:2014

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

packaging

package of 100 strips

technique(s)

microbe id | specific enzyme detection: suitable

antibiotic activity spectrum

Gram-negative bacteria

application(s)

agriculture
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Plesiomonas spp.
Aeromonas spp.
Neisseria spp.
Pseudomonas spp.

General description

Oxidase Strips are a rapid diagnostic test for the detection of the cytochrome oxidase activity of microorganisms. Oxidase Test is an important differential procedure which is performed on all gram-negative bacteria that are to be identified. In presence of the enzyme cytochrome oxidase (produced by gram-negative bacteria) the N,N-dimethyl-phenylenediamine, and alpha-naphthol impregnated on the strips react to indophenol blue which is visible by a color change.

Application

Oxidase Strips are used for detection of oxidase production by microorganisms like Neisseria, Alcaligenes, Aeromonas, Vibrio′s, Campylobacter and Pseudomonas.

Packaging

package of 100 test strips

Components

The kit contains plastic strips with a paper zone saturated with a solution of N,N-dimethyl-1,4-phenylene diamine and alpha-naphtol

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jake P Greenhalgh et al.
Food safety (Tokyo, Japan), 7(3), 74-78 (2020-01-31)
Edible insects present a potential solution to increasing global food insecurity. However, there is limited research on the microbial hazards they may pose. These include opportunistic pathogens like Cronobacter spp. (formerly Enterobacter sakazakii). In this study, nine types of ready-to-eat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service