Skip to Content
Merck
All Photos(1)

Documents

PHR1032

Supelco

Hydrochlorothiazide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

HCTZ, 6-Chloro-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8ClN3O4S2
CAS Number:
Molecular Weight:
297.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 186
traceable to Ph. Eur. H1200000
traceable to USP 1314009

API family

hydrochlorothiazide

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NS(=O)(=O)c1cc2c(NCNS2(=O)=O)cc1Cl

InChI

1S/C7H8ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-2,10-11H,3H2,(H2,9,12,13)

InChI key

JZUFKLXOESDKRF-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Hydrochlorothiazide is an antihypertensive and a diuretic that is used for the control of hypertension.

Application

Hydrochlorothiazide may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and spectrophotometry.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Carbonic anhydrase inhibitor
Carbonic anhydrase inhibitor

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB5026 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of enalapril maleate and hydrochlorothiazide in tablets by derivative UV spectrophotometry and high-performance liquid chromatography
Carlucci G, et al.
International Journal of Pharmaceutics, 93(1-3), 245-248 (1993)
Analysis of captopril and hydrochlorothiazide combination tablet formulations by liquid chromatography
Kirschbaum J and Perlman S
Journal of Pharmaceutical Sciences, 73(5), 686-687 (1984)
Simultaneous determination of benazepril hydrochloride and hydrochlorothiazide in tablets by second-order derivative spectrophotometry.
Panderi E I.
Journal of Pharmaceutical and Biomedical Analysis, 21(2), 257-265 (1999)
Simultaneous high-performance liquid chromatographic determination of telmisartan and hydrochlorothiazide in pharmaceutical preparation
Rane VP, et al.
Journal of Chromatographic Science, 46(10), 887-891 (2008)
A Dornhorst et al.
Lancet (London, England), 1(8421), 123-126 (1985-01-19)
14 hypertensive men with type II diabetes sequentially received, in random order, hydrochlorothiazide 50 mg twice a day, propranolol 80 mg twice a day, and both drugs in combination. The 3-week treatment periods were separated by a 1-week washout period.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service