Skip to Content
Merck

Skip To

91237

2-Propanol

greener alternative

analytical standard

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
UNSPSC Code:
12352104
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-661-7
Beilstein/REAXYS Number:
635639
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C), 44 mmHg ( 25 °C)

assay

≥99.9% (GC)

autoignition temp.

750 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

2.0-12.7 %, 93 °C

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.377 (lit.), n20/D 1.378

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble (completely)

density

0.785 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

greener alternative category

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

General description

2-Propanol is a secondary alcohol, which can find a wide number of applications as a solvent.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here
2-Propanol belongs to the class of volatile compounds and is an important aroma component found in wines, further adding to the organoleptic characteristic of the wines.

Application

2-Propanol may be used as an extraction solvent for the liquid-liquid extraction of acyclovir from human serum, which may be analyzed using high performance liquid chromatography(HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
2-Propanol may be used as an analytical reference standard for the quantification of the analyte in wine samples using gas chromatography techniques. It may also be used as a co-solvent for the supercritical carbon dioxide extraction of caffeine, chlorogenic acids and lipids from green coffee beans, analyzed by high-performance liquid chromatography technique.

Features and Benefits

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This solvent is safe to use and thus aligns with "Safer Solvents and Auxiliaries". Click here for more information.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
15704281096341.05008
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical

format

neat

format

neat

format

-

format

-

Quality Level

100

Quality Level

-

Quality Level

300

Quality Level

500

grade

analytical standard

grade

pharmaceutical primary standard

grade

ACS reagent

grade

-

assay

≥99.9% (GC)

assay

-

assay

≥99.8% (GC)

assay

≥99.8% (GC)


Still not finding the right product?

Explore all of our products under 2-Propanol


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters