Skip to Content
Merck
All Photos(1)

Key Documents

64139

Supelco

Ginsenoside Rg3

analytical standard

Synonym(s):

(3β,12β)-12,20-Dihydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, 20(S)-Ginsenoside-Rg3, Rg3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O13
CAS Number:
Molecular Weight:
785.01
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC[C@](C)(O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(C)(C)[C@@H]4CC[C@@]23C

InChI

1S/C42H72O13/c1-21(2)10-9-14-42(8,51)22-11-16-41(7)29(22)23(45)18-27-39(5)15-13-28(38(3,4)26(39)12-17-40(27,41)6)54-37-35(33(49)31(47)25(20-44)53-37)55-36-34(50)32(48)30(46)24(19-43)52-36/h10,22-37,43-51H,9,11-20H2,1-8H3/t22-,23+,24+,25+,26-,27+,28-,29-,30+,31+,32-,33-,34+,35+,36-,37-,39-,40+,41+,42-/m0/s1

InChI key

RWXIFXNRCLMQCD-JBVRGBGGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Kwang Kim et al.
Food chemistry, 141(2), 1369-1377 (2013-06-26)
The ginsenoside Rg3(S), which is one of the exceptional components of Korean red ginseng extract, has been known to have anti-cancer, anti-metastatic, and anti-obesity effects. An enzymatic bioconversion method was developed to obtain the ginsenoside Rg3(S) with a high specificity
J-P Liu et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3388-3396 (2012-07-24)
The purpose of this study is to investigate the potential subchronic toxicity of 20(S)-Ginsenoside Rg3(Rg3), by a 26-week repeated intramuscular administration in rats. Rg3 was administrated to rats at dose levels of 0, 4.2, 10.0 or 20.0 mg/kg/day. There was
Bo He et al.
Neuroscience letters, 526(2), 106-111 (2012-08-29)
Gensenosides, the active ingredients of Chinese herbal medicine Panax ginseng, have a wide spectrum of medical effects, such as anti-tumorigenic, angiosuppressive, adaptogenic, and anti-fatigue activities. In the present study, we have investigated the neuroprotective effect of 20(R)-ginsenoside Rg(3) (20(R)-Rg(3)) against
Xue-Yang Pan et al.
European journal of pharmacology, 683(1-3), 27-34 (2012-03-20)
Ginsenoside Rg3 (Rg3), one of the bioactive extracts found in ginseng root, was reported to have anti-cancer activity in various cancer models. The anti-proliferation effect of Rg3 on prostate cancer cells has been well reported. To test whether Rg3 has
Xiaojie Wei et al.
Fitoterapia, 83(4), 636-642 (2012-02-09)
Ginsenoside Rg3 is one of the active ingredients isolated from Panax ginseng C.A. Meyer. Previous investigation demonstrated that Rg3 was stereospecific in promotion of the immune response. The present study was designed to evaluate Rg3 and its epimers, 20(R)-Rg3 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service