Skip to Content
Merck
All Photos(2)

Key Documents

56805

Supelco

Ginsenoside Rh1

analytical standard

Synonym(s):

(3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl-β-D-glucopyranoside, Prosapogenin A2, Sanchinoside B2, Sanchinoside Rh1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H62O9
CAS Number:
Molecular Weight:
638.87
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C/CC[C@](C)(O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O)C(C)(C)[C@@H]4[C@H](C[C@@]23C)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O

InChI

1S/C36H62O9/c1-19(2)10-9-13-36(8,43)20-11-15-34(6)26(20)21(38)16-24-33(5)14-12-25(39)32(3,4)30(33)22(17-35(24,34)7)44-31-29(42)28(41)27(40)23(18-37)45-31/h10,20-31,37-43H,9,11-18H2,1-8H3/t20-,21+,22-,23+,24+,25-,26-,27+,28-,29+,30-,31+,33+,34+,35+,36-/m0/s1

InChI key

RAQNTCRNSXYLAH-RFCGZQMISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ji-Hae Yoon et al.
European journal of pharmacology, 679(1-3), 24-33 (2012-02-09)
Invasion and metastasis are the major causes of treatment failure in patients with cancer. Here, we investigated the effects of ginsenoside Rh1 on tumor invasion and metastasis in human hepatocellular carcinoma HepG2 cells and its possible mechanism of action. Rh1
Li Lai et al.
Planta medica, 75(8), 797-802 (2009-03-07)
20(S)-Ginsenoside Rh1 is one of the important protopanaxatriol ginsenosides and has been reported to be the main hydrolysis product reaching the systemic circulation after oral ingestion of ginseng. However, its pharmacokinetic characteristics and metabolic fate have never been reported. The
Hua Zheng et al.
International immunopharmacology, 11(4), 511-518 (2011-01-18)
In the present study, we examined the inhibitive effect of ginsenoside Rh1 on oxazolone-induced atopic dermatitis-like skin lesions in hairless mice. Oral administration of ginsenoside Rh1 improved clinical symptoms, and it was confirmed by histophathological analysis. In ginsenoside Rh1 (20mg/kg)
Wan Gu et al.
Biological & pharmaceutical bulletin, 36(1), 102-107 (2013-01-11)
Ginseng (the root of Panax ginseng C. A. MEYER), which contains protopanaxadiols and protopanaxatriols as its main constituents, has been used for many disorders, such as cancer, diabetes, inflammation, and hyperlipidemia. Of these ginsenosides, protopanaxadiol ginsenoside Rh2 alone is reported
Ji-Sun Jung et al.
Biochemical and biophysical research communications, 397(2), 323-328 (2010-06-01)
Microglial activation plays an important role in the pathogenesis of various neurodegenerative diseases by producing neurotoxic factors, such as pro-inflammatory cytokines and nitric oxide (NO). In the present study, we found that protopanaxatriol ginsenoside Rh1 suppresses NO, ROS, and TNF-alpha

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service