Skip to Content
Merck
All Photos(2)

Key Documents

08171

Supelco

Ginsenoside Rg2

analytical standard

Synonym(s):

(3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Chikusetsusaponin I, Panaxoside Rg2, Prosapogenin C2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O13
CAS Number:
Molecular Weight:
785.01
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥94% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2O[C@H]3C[C@]4(C)[C@H](C[C@@H](O)[C@@H]5[C@H](CC[C@@]45C)[C@@](C)(O)CC\C=C(/C)C)[C@@]6(C)CC[C@H](O)C(C)(C)[C@H]36)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C42H72O13/c1-20(2)11-10-14-42(9,51)22-12-16-40(7)28(22)23(44)17-26-39(6)15-13-27(45)38(4,5)35(39)24(18-41(26,40)8)53-37-34(32(49)30(47)25(19-43)54-37)55-36-33(50)31(48)29(46)21(3)52-36/h11,21-37,43-51H,10,12-19H2,1-9H3/t21-,22-,23+,24-,25+,26+,27-,28-,29-,30+,31+,32-,33+,34+,35-,36-,37+,39+,40+,41+,42-/m0/s1

InChI key

AGBCLJAHARWNLA-DQUQINEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginsenoside Rg2 is a saponin and a protopanaxatriol-type compound found in the root and stem leaves of ginseng.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T C Ma et al.
Arzneimittel-Forschung, 43(10), 1049-1052 (1993-10-01)
The effects of 20(S)-ginsenoside-Rg2 (GRg2, CAS 52286-74-5) and cyproheptadine (CYP, CAS 129-03-3) on acquisition, retention and retrieval were examined in male Wistar rats using a two-way active avoidance method. Learning and memory were estimated by the avoidance rate (%) and/or
Sung-Ryong Ko et al.
Chemical & pharmaceutical bulletin, 51(4), 404-408 (2003-04-04)
During investigation of the hydrolysis of a protopanaxatriol-type saponin mixture by various glycoside hydrolases, crude preparations of beta-galactosidase from Aspergillus oryzae and lactase from Penicillium sp. were found to produce two minor saponins, ginsenoside Rg(2) [6-O-(alpha-L-rhamnopyranosyl-(1-->2)-beta-D-glucopyranosyl)-20(S)-protopanaxatriol] and ginsenoside Rh(1) (6-O-beta-D-glucopyranosyl-20(S)-protopanaxatriol)
K Kudo et al.
European journal of pharmacology, 341(2-3), 139-144 (1998-05-23)
To investigate the relationship between the inhibitory effects of ginseng saponins (ginsenosides) on acetylcholine-evoked secretion of catecholamines and the structures of ginsenosides, we examined the effects of ginsenoside-Rg3 and -Rh2, which are panaxadiol saponins, 20(R)- and 20(S)-ginsenoside-Rg2, which are epimers
Fang-Jin Gui et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 850(1-2), 1-6 (2006-11-23)
20 (R,S)-Ginsenoside-Rg2, an anti-shock agent, is prescribed as a racemate. To analyze simultaneously the enantiomers of 20 (R)-ginsenoside-Rg2 and 20 (S)-ginsenoside-Rg2 in plasma, a simple and reproducible high-performance liquid chromatographic (HPLC) method has been developed. The enantiomeric separation and determination
E Tachikawa et al.
Biochemical pharmacology, 62(7), 943-951 (2001-09-07)
Since ginsenoside-Rg(3), one of the panaxadiol saponins isolated from the ginseng root, significantly inhibited the secretion of catecholamines from bovine adrenal chromaffin cells stimulated by acetylcholine (ACh), the properties of ginsenoside-Rg(3) inhibition were investigated. Although ginsenoside-Rg(3) inhibited the secretion evoked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service