Skip to Content
Merck
All Photos(2)

Key Documents

01518

Supelco

Ginsenoside Rd

analytical standard

Synonym(s):

(3β,12β)-20-(β-D-Glucopyranosyloxy)-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, Gypenoside VIII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H82O18
CAS Number:
Molecular Weight:
947.15
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C/CC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)C(C)(C)[C@@H]5CC[C@@]34C

InChI

1S/C48H82O18/c1-21(2)15-22(3)17-48(8,66-43-40(60)37(57)34(54)28(19-50)62-43)24-11-13-47(7)23-9-10-30-45(4,5)31(12-14-46(30,6)25(23)16-26(52)32(24)47)64-44-41(38(58)35(55)29(20-51)63-44)65-42-39(59)36(56)33(53)27(18-49)61-42/h15,22-44,49-60H,9-14,16-20H2,1-8H3/t22?,23?,24-,25?,26-,27+,28+,29+,30?,31?,32+,33+,34+,35+,36-,37-,38-,39+,40+,41+,42-,43-,44-,46+,47+,48-/m0/s1

InChI key

IRBVCFDYIGGFFY-ADSMOORZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginsenoside is a steroid-like molecular component having gonane steroid nucleus along with different sugar moieties attached to them found in the roots of Panax ginseng. Ginsenoside Rd is the main active component of ginsenosides, having the ability to inhibit Ca2+ influx through receptors and store-operated Ca2+ channel. From neural stem cells, it is seen to enhance astrocyte differentiation. It also reduces the 3-nitropropionic acid-induced motor impairment and cell loss in stratum. It constitutes to one of the major ingredient in total saponin content from Panax notoginseng.

Application

It was used as reference standard in UPLC/TOF-MS analysis of human plasma to determine Ginsenoside metabolites.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Li et al.
European journal of pharmacology, 652(1-3), 104-110 (2010-12-03)
Recently, it was revealed that the dysfunction of transmembrane Ca(2+) transport, results in an increase in intracellular Ca(2+)[Ca(2+)](i), which is involved in the process of atherosclerosis. We previously demonstrated that ginsenoside-Rd, a purified component from panax notoginseng, is a voltage-independent
X Liu et al.
European journal of neurology, 19(6), 855-863 (2012-01-12)
Ginsenoside-Rd is a receptor-operated calcium channel antagonist and has shown promise as a neuroprotectant in our phase II study. As an extended work, we sought to confirm its efficacy and safety of Ginsenoside-Rd in patients with acute ischaemic stroke. We
Li Wang et al.
The Journal of surgical research, 176(1), 267-274 (2011-08-30)
Organ transplantation is a life-saving procedure for patients with organ failure. However, the side effects of long-term application of classic immunosuppressant remain major obstacles for successful transplantation. Therefore, new and safe immunosuppressive drugs against acute and chronic rejection are eagerly
R Ye et al.
Neuroscience, 178, 169-180 (2011-01-12)
We previously found that ginsenoside Rd (Rd), one of the major active ingredients in Panax ginseng, protects neuronal cells from hydrogen peroxide and oxygen-glucose deprivation, an in vitro model of cerebral ischemia. In this study, we examined the protective effects
Ruidong Ye et al.
Neuropharmacology, 61(4), 815-824 (2011-06-15)
We previously found that ginsenoside Rd (Rd), one of the main active ingredients in Panax ginseng, protects against ischemic brain damage induced by oxygen-glucose deprivation in vitro and middle cerebral artery occlusion (MCAO) in vivo. Considering stroke happens frequently in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service