Skip to Content
Merck
All Photos(1)

Documents

63620

Millipore

D-(+)-Melezitose monohydrate

suitable for microbiology, ≥99.0%

Synonym(s):

Melezitose monohydrate, D-(+)-Melicitose monohydrate, α-D-(+)-Melicitose monohydrate, D-(+)-alpha-Melicitose monohydrate, D-(+)-3-O-α-D-Glucopyranosyl-D-glucose monohydrate, O-α-D-Glucopyranosyl-(1→3)-β-D-fructofuranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16 · H2O
CAS Number:
Molecular Weight:
522.45
Beilstein:
99539
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +88±2°, c = 4% in H2O

packaging

pkg of 10 g
pkg of 50 g

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Air sensitive.)

ign. residue

≤0.1% (as SO4)

color

colorless to white

mp

154-160 °C (dec.)

solubility

H2O: 50 mg/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

SMILES string

O.OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](CO)O[C@@]2(CO)O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H32O16.H2O/c19-1-5-8(23)11(26)13(28)16(30-5)32-15-10(25)7(3-21)33-18(15,4-22)34-17-14(29)12(27)9(24)6(2-20)31-17;/h5-17,19-29H,1-4H2;1H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15+,16-,17-,18+;/m1./s1

InChI key

CFXAVQUXSYFPDE-ZDVKUAOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Melezitose monohydrate is a crystalline form of the trisaccharide compound known as melezitose. Melezitose is composed of three glucose molecules linked together in a specific arrangement. It is commonly found in certain plants, such as cotton and buckwheat, as well as in the honeydew produced by some insect species. This compound can be used in various applications, including as a reference standard in analytical chemistry or as a component in research and development studies. It is also used in biochemical studies for investigating the metabolism and enzymatic activities related to sugars and carbohydrates.

Application

D-(+)-Melezitose monohydrate finds applications in various fields, including analytical chemistry, research and development, biochemical studies, and food industry. D-(+)-Melezitose can be used to identify clinical isolates of indole-positive and indole-negative Klebsiella spp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service