Skip to Content
Merck
All Photos(5)

Key Documents

M5885

Sigma-Aldrich

D-(+)-Maltose monohydrate

from potato, ≥99% (HPLC)

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
93798
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.28

biological source

potato

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

≤0.3% Glucose
≤1.0% maltotriose

color

white

useful pH range

5.0-7 (25 °C, 180 g/L)

mp

125.13 °C (1013.25 hPa)

solubility

180 g/L at 20 °C

application(s)

agriculture

storage temp.

room temp

SMILES string

O.OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fecal microbial and metabolic characteristics of swine from birth to market.: This research analyzes the microbial and metabolic profiles of swine at various life stages. ᴅ-(+)-Maltose is utilized in the culture media to study microbial growth, providing insights into the gut microbiota of livestock and its implications for animal health and productivity (He et al., 2023).

  • Quantification of disaccharides in solution using isomer-selective ultraviolet photodissociation of hydrogen-bonded clusters in the gas phase.: This study presents a novel method for quantifying disaccharides, including ᴅ-(+)-Maltose, using ultraviolet photodissociation. The method provides precise measurements of disaccharide concentrations, beneficial for chemists involved in analytical biochemistry (Nagai et al., 2023).


Biochem/physiol Actions

Maltose is a disaccharide containing two glucose molecules with an α(1→4) glycosidic linkage. Maltose can be derived from starch in food through the action of amylase. Maltose can be found in many food products, including beer, cereals, and pasta.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiajie Lian et al.
Bio-protocol, 8(8), e2810-e2810 (2018-04-20)
Botrytis cinerea (B. cinerea) attacks many crops of economic importance, represents one of the most extensively studied necrotrophic pathogens. Inoculation of B. cinerea and phenotypic analysis of plant resistance are key procedures to investigate the mechanism of plant immunity. Here
Salma Alavi et al.
STAR protocols, 1(3), 100200-100200 (2020-12-31)
The gut microbiome plays an important role in the exclusion of pathogens and, thus, infection outcomes. Microbiome-pathogen interaction studies are complicated by a lack of tractable animal models and differences in animal model versus human microbiomes. We have adapted the
Katharina Isensee et al.
The Analyst, 143(24), 6025-6036 (2018-11-08)
Continuous glucose monitoring enables an improved disease management for people with diabetes. However, state-of-the-art, enzyme-based, minimally invasive sensors lose their sensitivity over time and have to be replaced periodically. Here, we present the in vitro investigation of a quantum cascade
Camila Gabriel Kato-Schwartz et al.
Food research international (Ottawa, Ont.), 137, 109462-109462 (2020-11-26)
A practical approach to control glycemia in diabetes is to use plant natural products that delay hydrolysis of complex sugars and promote the diminution of the release of glucosyl units into the blood plasma. Polyphenolics have been described as being
Paul M Airs et al.
Acta tropica, 191, 221-227 (2019-01-12)
Toxic Sugar Baits (TSBs) are an inexpensive and field-applicable approach to deliver a variety of insecticides to sugar-seeking mosquitoes. We reasoned that carbohydrate chemistry could alter the performance and efficacy of TSBs. In this study, the uptake, distribution, and survival

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service