Skip to Content
Merck
All Photos(1)

Key Documents

90210

Millipore

D-(+)-Trehalose dihydrate

suitable for microbiology, ≥99.0%, composed of two α-glucose units that is used as a protectant, stabilizer and to support proper folding

Synonym(s):

Trehalose dihydrate, a-d-glucopyranosyl-a-d-glucopyranoside, α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Beilstein:
5322018
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +176 to +186°, c = 2% in H2O

mol wt

378.33  g/mol

impurities

≤0.1% glucose

ign. residue

≤0.1% (as SO4)

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

application(s)

microbiology

SMILES string

[H]O[H].[H]O[H].OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

InChI key

DPVHGFAJLZWDOC-PVXXTIHASA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Trehalose is a disaccharide composed of two α-glucose units. It is used as a cryoprotectant in a variety of cell freezing media. Used as a protectant in transportation media. It predominantly exists among the species that support anhydrobiosis (life without water). It serves as a carbohydrate reserve in microorganisms and protects them from adverse conditions, such as heat, cold, dehydration, desiccation, and oxidation. Trehalose acts as a chemical chaperone affecting protein folding. It is known to prevent the generation of amyloids and accumulation of β-amyloid in vitro. A similar effect of trehalose has been observed with Huntingtin protein aggregation. Thus, it might serve as a protective agent in Alzheimer′s disease and Huntington′s disease. Trehalose stimulates the better initial growth of Salmonella cells.

Application

D-(+)-Trehalose can be used as a carbohydrate reserve in certain cell cultures to tide over environmental stresses. It can also be used in biopharmaceutical monoclonal antibody formulations and as a protein stabilizer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwani Kesarwani et al.
Scientific reports, 11(1), 3049-3049 (2021-02-06)
Due to the limited utility of Bacillus Calmette-Guérin (BCG), the only approved vaccine available for tuberculosis, there is a need to develop a more effective and safe vaccine. We evaluated the safety and efficacy of a dry powder aerosol (DPA)
Long Zheng et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(14), 3694-3706 (2020-04-11)
The murine Lym-1 mAb targets a discontinuous epitope (Lym-1 epitope) on several subtypes of HLA-DR, which is upregulated in a majority of human B-cell lymphomas and leukemias. Unlike CD19, the Lym-1 epitope does not downregulate upon crosslinking, which may provide
Xiu He et al.
Cells, 9(1) (2020-01-18)
Silicosis is an occupational lung disease characterized by persistent inflammation and irreversible fibrosis. Crystalline silica (CS) particles are mainly phagocytized by alveolar macrophages (AMs), which trigger apoptosis, inflammation, and pulmonary fibrosis. Previously, we found that autophagy-lysosomal system dysfunction in AMs
Cristina Blázquez et al.
eLife, 9 (2020-08-11)
The use of cannabis is rapidly expanding worldwide. Thus, innovative studies aimed to identify, understand and potentially reduce cannabis-evoked harms are warranted. Here, we found that Δ9-tetrahydrocannabinol, the psychoactive ingredient of cannabis, disrupts autophagy selectively in the striatum, a brain
Sebastian D Burz et al.
Scientific reports, 9(1), 8897-8897 (2019-06-22)
Owing to the growing recognition of the gut microbiota as a main partner of human health, we are expecting that the number of indications for fecal microbiota transplantation (FMT) will increase. Thus, there is an urgent need for standardization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service