Skip to Content
Merck
All Photos(1)

Key Documents

63630

Millipore

D-(+)-Melibiose

suitable for microbiology, ≥99.0%

Synonym(s):

Raffinose, Melizitose, α-D-Galactosyl-(1→6)-α-D-glucopyranoside, Galactosyl D-glucose, Galactosylglucose, D-Galactopyranosyl-(1→6)-D-glucose, Melibiose, Galactinol, D-(+)-Galactosyl-(1→6)-D-(+)-glucose, α-D-Gal(1→6)β-D-Glc, 6-O-α-D-Galactopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93360
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.74

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +137±3°, 10 hr, c = 5% in H2O

packaging

pkg of 10 g
pkg of 50 g

ign. residue

≤0.1% (as SO4)

color

colorless to white

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

SMILES string

OC[C@H]1O[C@H](OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h1,4-12,14-21H,2-3H2/t4-,5+,6+,7+,8+,9-,10-,11+,12-/m0/s1

InChI key

AYRXSINWFIIFAE-GFRRCQKTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Melibiose is a reducing disaccharide composed of a unit of galactose linked to a unit of glucose by a glycosidic bond α (1 → 6). It is commonly found in plants and the roots of certain legumes. D-(+)-Melibiose is not metabolized by humans but serves as a substrate for certain bacteria and fungi. It is used as a differential medium component in microbiological media to detect the fermentation of melibiose by microorganisms. It helps identify specific bacterial species based on their ability to ferment melibiose.

Application

D-(+)-Melibiose has applications in the fields of microbiology, research and development, and in food and pharmaceutical industries. Melibiose fermentation test is used to differentiate bacteria like Enterobacteriaceae (like Salmonella, E.coli, shigella, Citrobacter, Klebsiella, Enterobacter) and yeasts. It is also used for the differentiation between the plant pathogenic bacteria Klebsiella pneumoniae and K. variicola.

Storage and Stability

Keep the container tightly closed in a dry and well-ventilated place.Hygroscopic, Store under inert gas.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saswati Chakladar et al.
Biochemistry, 50(20), 4298-4308 (2011-04-19)
The MelA gene from Citrobacter freundii, which encodes a glycosyl hydrolase family 4 (GH4) α-galactosidase, has been cloned and expressed in Escherichia coli. The recombinant enzyme catalyzes the hydrolysis of phenyl α-galactosides via a redox elimination-addition mechanism involving oxidation of
Jennifer L Chaytor et al.
Glycobiology, 22(1), 123-133 (2011-08-20)
The ice recrystallization inhibition activity of various mono- and disaccharides has been correlated with their ability to cryopreserve human cell lines at various concentrations. Cell viabilities after cryopreservation were compared with control experiments where cells were cryopreserved with dimethylsulfoxide (DMSO).
S Vivek Jakkula et al.
Journal of bacteriology, 194(20), 5538-5544 (2012-08-07)
The melibiose permease of Salmonella enterica serovar Typhimurium (MelB(St)) catalyzes symport of melibiose with Na(+), Li(+), or H(+). Bioinformatics and mutational analyses indicate that a conserved Gly117 (helix IV) is a component of the Na(+)-binding site. In this study, Gly117
Peng Lin et al.
Acta biochimica et biophysica Sinica, 40(12), 1029-1038 (2008-12-18)
A dimeric 50 kDa melibiose-binding lectin was isolated from the seeds of the cultivar of soybean (Glycine max), called the small glossy black soybean. The isolation procedure comprised ion exchange chromatography on Q Sepharose, SP Sepharose and Mono Q followed
Meritxell Granell et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(51), 22078-22083 (2010-12-08)
The melibiose carrier from Escherichia coli (MelB) couples the accumulation of the disaccharide melibiose to the downhill entry of H(+), Na(+), or Li(+). In this work, substrate-induced FTIR difference spectroscopy was used in combination with fluorescence spectroscopy to quantitatively compare

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service