Skip to Content
Merck
All Photos(1)

Key Documents

46148

Supelco

Corticosterone

VETRANAL®, analytical standard

Synonym(s):

11β,21-Dihydroxy-4-pregnene-3,20-dione, 11β,21-Dihydroxyprogesterone, 4-Pregnene-11β,21-diol-3,20-dione, Kendall’s Compound B, Reichstein’s Substance H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
Beilstein:
2339601
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

179-183 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO

InChI

1S/C21H30O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,14-17,19,22,24H,3-8,10-11H2,1-2H3/t14-,15-,16+,17-,19+,20-,21-/m0/s1

InChI key

OMFXVFTZEKFJBZ-HJTSIMOOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Corticosterone is one of the principal steroids secreted by the rat adrenal gland and found in rat plasma.[1][2]

Application

Corticosterone may be used as a reference standard in the determination of corticosterone in plasma samples using high-performance liquid chromatography (HPLC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of plasma corticosterone using high-performance liquid chromatography.
Woodward HJC and Emery WP
Journal of Chromatography. B, Biomedical Sciences and Applications, 419(2), 280-284 (1987)
Selective retention of corticosterone by limbic structures in rat brain.
McEwen SB, et al.
Nature, 220(5170), 911-911 (1968)
The estimation of small amounts of corticosterone in rat plasma.
Zenker N and Bernstein ED
The Journal of Biological Chemistry, 231(2), 695-701 (1958)
Damiano Azzinnari et al.
Neuropharmacology, 85, 328-341 (2014-06-08)
In neuropsychiatry, animal studies demonstrating causal effects of environmental manipulations relevant to human aetiology on behaviours relevant to human psychopathologies are valuable. Such valid models can improve understanding of aetio-pathophysiology and preclinical discovery and development of new treatments. In depression
E N de Haas et al.
Poultry science, 92(9), 2259-2269 (2013-08-21)
Little is known about the relationship between welfare traits and production in laying hen parent stock (PS). In commercial laying hens and pure lines, it is known that aspects associated with reduced welfare such as high fear, stress, and feather

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service