Skip to Content
Merck
All Photos(2)

Key Documents

P9521

Sigma-Aldrich

21-Deoxycortisol

Synonym(s):

11β,17α-Dihydroxy-4-pregnene-3,20-dione, 11β,17α-Dihydroxyprogesterone, 21-Desoxycortisol, 4-Pregnene-11β,17α-diol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98.00% (TLC)

form

powder

solubility

ethanol: 9.80-10.20 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

InChI

1S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h10,15-18,24-25H,4-9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1

InChI key

LCZBQMKVFQNSJR-UJPCIWJBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

21-Deoxycortisol is a 17-hydroxyprogesterone derivative. It serves as a marker for congenital adrenal hyperplasia, an autosomal recessive disorder, indicated by 21-hydroxylase deficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Superior discriminating value of ACTH-stimulated serum 21-deoxycortisol in identifying heterozygote carriers for 21-hydroxylase deficiency
Costa BFA, et al.
Clin. Endocrinol., 73(6), 700-706 (2010)
Radha Chaube et al.
Theriogenology, 105, 90-96 (2017-09-25)
In the present study, distribution of steroid hormones (estradiol-17β (E
Cristina A Martinez et al.
Genes, 11(8) (2020-08-06)
Anxiety, chronical stress, and depression during pregnancy are considered to affect the offspring, presumably through placental dysregulation. We have studied the term placentae of pregnancies clinically monitored with the Beck's Anxiety Inventory (BAI) and Edinburgh Postnatal Depression Scale (EPDS). A
J Fiet et al.
Annals of clinical biochemistry, 31 ( Pt 1), 56-64 (1994-01-01)
21-deoxycortisol (21-DF) is a steroid of strictly adrenal origin formed by the 11-hydroxylation of 17-hydroxyprogesterone. This metabolic pathway is minor in normal subjects, in whom basal plasma concentrations range from 0.03 to 0.63 nmol/L and from 0.865 to 1.50 nmol/L
M Hill et al.
Steroids, 60(9), 615-620 (1995-09-01)
A radioimmunoassay of three deoxycorticoids, namely 11 beta,17 alpha-dihydroxy-4-pregnene-3,20-dione (21-deoxycortisol), 17 alpha,21-dihydroxy-4-pregnene-3,20-dione (11-deoxycortisol), and 21-hydroxy-4-pregnene-3,20-dione (11-deoxycorticosterone) which are important for differential diagnosis of congenital adrenal disorders, is described and evaluated. Antisera against 3-(O-carboxymethyl)oximes conjugated to bovine serum albumin were raised

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service