Skip to Content
Merck
All Photos(1)

Documents

69673

Supelco

Homovanillic acid

analytical standard

Synonym(s):

4-Hydroxy-3-methoxyphenylacetic acid, HVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)CH2CO2H
CAS Number:
Molecular Weight:
182.17
Beilstein:
2213447
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

142-145 °C (lit.)
142-145 °C

application(s)

pharmaceutical

format

neat

SMILES string

COc1cc(CC(O)=O)ccc1O

InChI

1S/C9H10O4/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4,10H,5H2,1H3,(H,11,12)

InChI key

QRMZSPFSDQBLIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Homovanillic acid may be used as a reference standard in the determination of homovanillic acid in samples of human cerebrospinal fluid using gas chromatography coupled with electron capture detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the occurrence of homovanillic acid in brain and cerebrospinal fluid and its determination by a fluorometric method.
Sjoquist B and Anggard E.
Life Sciences, 2(7), 448-458 (1963)
Xiaoli Liu et al.
Neurotoxicity research, 38(1), 27-37 (2020-03-22)
Parkinson's disease (PD) is a neurodegenerative disorder characterized by the accumulation of Lewy bodies and loss of dopaminergic neurons in the substantia nigra pars compacta (SNpC). MPTP is widely used to generate murine PD model. In addition to classical motor
Kyung Tai Lee et al.
Foot & ankle international, 35(12), 1329-1333 (2014-09-23)
Hallux valgus is speculated to increase the load on the second metatarsophalangeal (MTP) joint, possibly inducing degenerative osteoarthritis. In addition, the severity of arthritis may be correlated with the severity of hallux valgus. This study evaluated the association of arthritis
Kangwei Li et al.
Journal of ethnopharmacology, 250, 112487-112487 (2019-12-21)
Seahorses (Hippocampus erectus), belonging to syngnathidae of syngnathiformes, are a traditional Chinese medicine for increasing and balancing vital energy within the body and brain, as well as calming mood and improving sleep. Based on the hypothesis of monoamine neurotransmitter deficiency
M A Aguilar et al.
Brain research bulletin, 113, 27-33 (2015-03-01)
Although the consumption of cocaine is frequent in young users of MDMA (3,4-methylenedioxymethamphetamine), the influence of exposure to cocaine on the rewarding effects of MDMA in adolescents has not been studied. The purpose of the present work was to evaluate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service