Skip to Content
Merck
All Photos(3)

Documents

457116

Sigma-Aldrich

Decane

anhydrous, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28
Beilstein:
1696981
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.06

grade

anhydrous

Quality Level

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

410 °F

expl. lim.

2.6 %

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 1.411 (lit.)

bp

174 °C (lit.)

mp

−30 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decane (C10 H12) is an acyclic saturated hydrocarbon.

Application

Decane has been used as an organic solvent for nanocrystal suspension.

Biochem/physiol Actions

Decane-1,2-diol derivative might prevent cancer development.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chemistry, An Introduction to General, Organic, and Biological Chemistry (2016)
Optical Properties of CdSe/ZnS Nanocrystals
Gaigalas AK, et al.
Journal of research of the national institute of standards and technology, (119), 610-628 (2014)
Brocilo D, et al.
IEEE International Conference on Dielectric Liquids (ICDL). 2011 null
Hyunjoong Chung et al.
Nature communications, 9(1), 278-278 (2018-01-20)
Martensitic transition is a solid-state phase transition involving cooperative movement of atoms, mostly studied in metallurgy. The main characteristics are low transition barrier, ultrafast kinetics, and structural reversibility. They are rarely observed in molecular crystals, and hence the origin and
High Pressure Oxidation of Liquid Fuels from Low to High Temperature. 3. n-Decane.
Dagaut P, et al.
Combustion Science and Technology, 103(1-6), 349-359 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service