Skip to Content
Merck
All Photos(1)

Key Documents

172456

Sigma-Aldrich

Tetradecane

≥99%

Synonym(s):

n-Tetradecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12CH3
CAS Number:
Molecular Weight:
198.39
Beilstein:
1733859
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010124
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.83 (vs air)

vapor pressure

1 mmHg ( 76.4 °C)

Assay

≥99%

autoignition temp.

455 °F

refractive index

n20/D 1.429 (lit.)

bp

252-254 °C (lit.)

mp

5.5 °C (lit.)

density

0.762 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCC

InChI

1S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3

InChI key

BGHCVCJVXZWKCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tetradecane is an alkane hydrocarbon containing 14 carbon atoms. It is generally used as an organic solvent in synthesis Nonpolar aliphatic solvent, n-tetradecane, is good for aliphatic side chains, but poor for aromatic backbones.

Application

Tetradecane was used to study the thermodiffusion behavior of the microemulsion droplets of H2O/n-alkanes/C12E5 (pentaethylene glycol monododecyl ether).

Tetradecane is also used as:

  • A core material in the preparation of NEPCMs(nano-encapsulated phase change materials) along with sodium dodecyl sulfate as emulsifier, resorcin as system modifier, and urea–formaldehyde resin as the shell material.
  • A selective solvent to study the phase behavior of block copolymer melts.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and surface activities of organic solvent-soluble fluorinated surfactants
Li GL, et al.
Journal of Fluorine Chemistry, 130(7), 674-681 (2009)
Preparation and characterization of nano-encapsulated n-tetradecane as phase change material for thermal energy storage
Fang G, et al.
Chemical Engineering Journal, 153(1-3), 217-221 (2009)
Theano Mikrou et al.
Molecules (Basel, Switzerland), 25(17) (2020-08-26)
Extra virgin olive oil (EVOO) is an important component of the Mediterranean diet and a highly priced product. Despite the strict legislation to protect it from fraudulent practices, there is an increasing demand to characterize EVOOs and evaluate their authenticity.
Marco Dompé et al.
Polymers, 12(2) (2020-02-09)
Underwater adhesion represents a huge technological challenge as the presence of water compromises the performance of most commercially available adhesives. Inspired by natural organisms, we have designed an adhesive based on complex coacervation, a liquid-liquid phase separation phenomenon. A complex
Christoph Aeppli et al.
Environmental science & technology, 43(23), 8813-8820 (2009-12-01)
Mass transfer of organic contaminants from nonaqueous phase liquids to the aqueous phase can significantly modulate the observable carbon isotope fractionation behavior associated with contaminant transformation. We evaluated the effects of kinetic interphase mass transfer between tetradecane and water on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service