Skip to Content
Merck
All Photos(3)

Documents

D901

Sigma-Aldrich

Decane

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28
Beilstein:
1696981
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.06

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

410 °F

expl. lim.

2.6 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.411 (lit.)

bp

174 °C (lit.)

mp

−30 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decane (n-Decane), an aliphatic hydrocarbon, is a reference fuel. Its oxidation over a range of temperature has been studied. Its crystal structure has been investigated at 150K. The cracking of decane can be performed using electrohydrodynamic atomization system (EHDA).

Application

Decane has been used as a solvent in the preparation of high alkane phase emulsified suspensions (HAPES). It may also be used as a solvent to suspend the CdSe/ZnS nanocrystals during absorbance measurement.
Internal standard in the GC analysis of oxalic, malonic, and succinic acids in biological materials.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Colored ceramic foams with tailored pore size and surface functionalization used as spawning plates for fish breeding.
Kroll S, et al.
Ceramics International, 40(10), 15763-15773 (2014)
Brocilo D, et al.
IEEE International Conference on Dielectric Liquids (ICDL). 2011 null
High Pressure Oxidation of Liquid Fuels from Low to High Temperature. 3. n-Decane.
Dagaut P, et al.
Combustion Science and Technology, 103(1-6), 349-359 (1994)
n-Decane.
Bond AD and Davies JE.
Acta Crystallographica Section E, Structure Reports Online, 58(2), 196-197 (2002)
Optical Properties of CdSe/ZnS Nanocrystals.
Gaigalas AK, et al.
Journal of research of the national institute of standards and technology, 119, 610-628 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service