Skip to Content
Merck
All Photos(1)

Key Documents

556432

Sigma-Aldrich

4-Ethynyl-α,α,α-trifluorotoluene

97%

Synonym(s):

1-Ethynyl-4-(trifluoromethyl)benzen, 1-Ethynyl-4-(trifluoromethyl)benzene, 1-Trifluoromethyl-4-ethynylbenzene, 4-(Trifluoromethyl)phenylacetylene, 4-Ethynyl(trifluoromethyl)benzene, 4-Ethynyl-trifluorotoluene, 4-Trifluorophenylacetylene, [4-(Trifluoromethyl)phenyl]ethyne, p-(Trifluoromethyl)phenylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4CF3
CAS Number:
Molecular Weight:
170.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.4650 (lit.)

bp

78-80 °C/2 mmHg (lit.)

density

1.043 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)c1ccc(cc1)C#C

InChI

1S/C9H5F3/c1-2-7-3-5-8(6-4-7)9(10,11)12/h1,3-6H

InChI key

XTKBMZQCDBHHKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Ethynyl-α,α,α-trifluorotoluene reacts with ethanol to afford the corresponding α,β-alkynyl ester. 4-Ethynyl-α,α,α-trifluorotoluene plays the role of a functionalized alkyne in copper-catalyzed azide-alkyne cycloaddition (CuAAC) reaction on BrPP thin films.

Application

4-Ethynyl-α,α,α-trifluorotoluene may be used to synthesize the following:
  • 6,13-bis(4-trifluoromethylphenylethynyl)pentacene
  • 1,2-dialkynylimidazoles
  • trans-[Co(cyclam)(p-CCC6H4CF3)2]OTf complex (where cyclam - 1,4,8,11-tetraazacyclotetradecane; 4-ethynyl-α,α,α-trifluorotoluene - p-CCC6H4CF3; OTf- trifluoromethane sulfonate)
  • 3-spiroazetidinimine-2-oxindoles
Used to synthesize self-assembled monolayers (SAMs).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.1 °F - closed cup

Flash Point(C)

20.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thakker U P, et al.
Inorgorganica Chimica Acta, 411, 158- 164 (2014)
Journal of Polymer Science Part A: Polymer Chemistry, 42, 541-550 (2004)
Synthesis of α, β-Alkynyl Esters and Unsymmetrical Maleate Esters Catalyzed by Pd/C; An Efficient Phosphine-Free Catalytic System for Oxidative Alkoxycarbonylation of Terminal Alkynes.
Gadge ST and Bhanage BM.
Synlett, 24(8), 981-986 (2013)
Rodney T Chen et al.
Langmuir : the ACS journal of surfaces and colloids, 26(5), 3388-3393 (2009-11-12)
A brominated plasma polymer (BrPP) thin film was fabricated on a variety of substrate surfaces (silicon wafers, glass, gold, and polymers) via the radio frequency glow discharge of 1-bromopropane. This BrPP thin film was highly adherent and stable and was
"5H-Cyclopentapyrazines from 1, 2-Dialkynylimidazoles"
Kerwin.MS and Nadipuram A
Synlett, 2004(08), 1404-1408 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service