Skip to Content
Merck
All Photos(1)

Key Documents

45371

Supelco

Carboxine

PESTANAL®, analytical standard

Synonym(s):

5,6-Dihydro-2-methyl-N-phenyl-1,4-oxathiin-3-carboxamide, Carbathiine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO2S
CAS Number:
Molecular Weight:
235.30
Beilstein:
983249
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1=C(SCCO1)C(=O)Nc2ccccc2

InChI

1S/C12H13NO2S/c1-9-11(16-8-7-15-9)12(14)13-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3,(H,13,14)

InChI key

GYSSRZJIHXQEHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y N Yin et al.
Phytopathology, 101(8), 986-995 (2011-04-08)
Botrytis cinerea isolates obtained from apple orchards were screened for resistance to boscalid. Boscalid-resistant (BosR) isolates were classified into four phenotypes based on the levels of the concentration that inhibited fungal growth by 50% relative to control. Of the 220
Marina DellaGreca et al.
Journal of agricultural and food chemistry, 52(20), 6228-6232 (2004-09-30)
Sunlight exposure of aqueous suspensions of carboxin (1) causes its phototransformation to sulfoxide 2 and minor components. Similar effects are observed in the presence of humic acid or nitrate or at different pH values. Photoproducts 2-9 were isolated by chromatographic
Akinori Ando et al.
Current genetics, 55(3), 349-356 (2009-05-26)
The sdhB gene encoding an iron-sulfur (Ip) subunit of succinate dehydrogenase (SDH, EC 1.3.99.1) complex was cloned from Mortierella alpina 1S-4. The deduced amino acid sequence of SdhB from M. alpina 1S-4 showed high similarity to those of SdhB from
Yanshen Deng et al.
Toxicon : official journal of the International Society on Toxinology, 42(4), 351-357 (2003-09-25)
The effect of surangin B, an insecticidal natural product coumarin, on presynaptic release of endogenous amino acids was investigated using a purified synaptosomal fraction isolated from mouse brain. Surangin B stimulated the release of glutamic acid (GLU), gamma-aminobutyric acid (GABA)
Yoko Shima et al.
Fungal genetics and biology : FG & B, 46(1), 67-76 (2008-11-11)
Mutants exhibiting resistance to the fungicide, carboxin, were isolated from Aspergillus oryzae, and the mutations in the three gene loci, which encode succinate dehydrogenase (SDH) B, C, and D subunits, were identified to be independently responsible for the resistance. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service