Skip to Content
Merck
All Photos(1)

Key Documents

45463

Supelco

Diuron

PESTANAL®, analytical standard

Synonym(s):

3-(3,4-Dichlorophenyl)-1,1-dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10Cl2N2O
CAS Number:
Molecular Weight:
233.09
Beilstein:
2215168
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CN(C)C(=O)Nc1ccc(Cl)c(Cl)c1

InChI

1S/C9H10Cl2N2O/c1-13(2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5H,1-2H3,(H,12,14)

InChI key

XMTQQYYKAHVGBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Giacomazzi et al.
Chemosphere, 56(11), 1021-1032 (2004-07-28)
Diuron is a biologically active pollutant present in soil, water and sediments. A synthesis of literature data on its physicochemical properties, partitioning behaviour, abiotic and biotic transformations, toxicological and ecotoxicological impacts has been here performed. Data have shown that diuron
Ezequiel Petrillo et al.
Science (New York, N.Y.), 344(6182), 427-430 (2014-04-26)
Light is a source of energy and also a regulator of plant physiological adaptations. We show here that light/dark conditions affect alternative splicing of a subset of Arabidopsis genes preferentially encoding proteins involved in RNA processing. The effect requires functional
Lijin Tian et al.
Physical chemistry chemical physics : PCCP, 15(9), 3146-3154 (2013-01-24)
Photosystems I (PSI) and II (PSII) are two major pigment-protein complexes of photosynthetic organisms that function in series to convert sunlight energy into chemical energy. We have studied the picosecond fluorescence behaviour of the core of both photosystems in vivo
Tobias Wunder et al.
Planta, 237(2), 541-558 (2012-10-23)
Reversible phosphorylation of LHCII, the light-harvesting complex of photosystem II, controls its migration between the two photosystems (state transitions), and serves to adapt the photosynthetic machinery of plants and green algae to short-term changes in ambient light conditions. The thylakoid
Sheridan M Martin et al.
Journal of hazardous materials, 231-232, 70-78 (2012-07-17)
We studied the sorption-desorption behaviour of two herbicides (diuron and artrazine) in a soil rich in Fe and Al oxides (Ferrosol), either amended freshly with two different types of biochars or that contained biochars aged under field conditions. Standard batch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service